PA1-27843 detects G3BP from human, mouse, and rat samples.
PA1-27843 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.
The immunogen is a synthetic peptide corresponding to human Ras-GTPase-activating protein.
This gene encodes one of the DNA-unwinding enzymes which prefers partially unwound 3'-tailed substrates and can also unwind partial RNA/DNA and RNA/RNA duplexes in an ATP-dependent fashion. This enzyme is a member of the heterogeneous nuclear RNA-binding proteins and is also an element of the Ras signal transduction pathway. It binds specifically to the Ras-GTPase-activating protein by associating with its SH3 domain. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.