Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GABBR1 Antibodies

          Invitrogen

          GABBR1 Polyclonal Antibody

          View all (27) GABBR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite GABBR1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          GABBR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          GABBR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GABBR1 Antibody (PA5-33566) in IHC (P)

          Immunohistochemistry analysis of GABBR1 in human Brain, Glioblastoma. Samples were incubated with GABBR1 polyclonal antibody (Product # PA5-33566). Formalin-fixed, paraffin-embedded tissue after heat-induced antigen retrieval. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GABBR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GABBR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GABBR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GABBR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GABBR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GABBR1 Polyclonal Antibody

          Product Details

          PA5-33566

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          9-18 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bovine, Hamster, Human, Mouse, Non-human primate, Pig, Rabbit, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic 15 amino acid peptide from N-terminal extracellular domain of human GABBR1.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.1% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2550946

          Product Specific Information

          Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Monkey, Mouse, Rat, Hamster, Elephant, Panda, Bovine, Rabbit, Pig, Opossum (100%).

          Target Information

          Receptor for GABA. The activity of this receptor is mediated by G-proteins that inhibit adenylyl cyclase activity, stimulates phospholipase A2, activates potassium channels, inactivates voltage-dependent calcium-channels and modulates inositol phospholipids hydrolysis. Plays a critical role in the fine-tuning of inhibitory synaptic transmission. Pre-synaptic GABA-B-R inhibit neurotransmitter release by down-regulating high-voltage activated calcium channels, whereas postsynaptic GABA-B-R decrease neuronal excitability by activating a prominent inwardly rectifying potassium (Kir) conductance that underlies the late inhibitory postsynaptic potentials. Not only implicated in synaptic inhibition but also in hippocampal long-term potentiation, slow wave sleep, muscle relaxation and antinociception.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: bM573K1.1.1 (gamma-aminobutyric acid (GABA) B receptor, 1a); DAAP-188P13.3; dJ271M21.1.1; dJ271M21.1.2; FLJ92613; G-protein coupled receptor; GABA(B) receptor; GABA-B receptor 1; GABA-B receptor, R1 subunit; GABA-B-R1; Gaba-b-r1a; Gaba-b1; GABA-BR1; Gaba-br1a receptor; GABA-BR1b receptor; GABAB; GABABR1; Gababr1 receptor; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 1; gamma-aminobutyric acid (GABA) B receptor, 1; gamma-aminobutyric acid (GABA-B) receptor, 1; Gamma-aminobutyric acid type B receptor subunit 1; gb1; GBR1; HGB1a; metabotropic GABA receptor; OTTHUMP00000109099; OTTHUMP00000214725; seven transmembrane helix receptor; unnamed protein product

          View more View less

          Gene Aliases: bM573K1.1; GABAB1; GABABR1; GABBR1; GABBR1-3; GB1; GPRC3A; I79_015084; NEDLC

          View more View less

          UniProt ID: (Human) Q9UBS5, (Mouse) Q9WV18, (Rat) Q9Z0U4

          View more View less

          Entrez Gene ID: (Pig) 100141417, (Rabbit) 100353026, (Human) 2550, (Bovine) 513785, (Chinese hamster) 100770643, (Mouse) 54393, (Rat) 81657

          View more View less

          Function(s)
          G-protein coupled receptor activity G-protein coupled GABA receptor activity protein binding GABA receptor activity protein heterodimerization activity G protein-coupled neurotransmitter receptor activity involved in regulation of postsynaptic membrane potential G protein-coupled neurotransmitter receptor activity involved in regulation of presynaptic membrane potential extracellular matrix protein binding signal transducer activity transcription factor binding G-protein coupled receptor
          Process(es)
          osteoblast differentiation signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway gamma-aminobutyric acid signaling pathway negative regulation of cell proliferation regulation of glutamate secretion positive regulation of glutamate secretion negative regulation of gamma-aminobutyric acid secretion negative regulation of epinephrine secretion negative regulation of dopamine secretion response to nicotine response to ethanol negative regulation of synaptic transmission synaptic transmission, GABAergic regulation of postsynaptic membrane potential positive regulation of growth hormone secretion regulation of presynaptic membrane potential neuron-glial cell signaling negative regulation of adenylate cyclase activity regulation of cAMP biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0