This antibody is specific for the ~52 kDa d-subunit of the GABA-A receptor.
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel.