Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes the alpha subunit of glucosidase II and a member of the glycosyl hydrolase 31 family of proteins. The heterodimeric enzyme glucosidase II plays a role in protein folding and quality control by cleaving glucose residues from immature glycoproteins in the endoplasmic reticulum. Expression of the encoded protein is elevated in lung tumor tissue and in response to UV irradiation. Mutations in this gene cause autosomal-dominant polycystic kidney and liver disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-glucosidase 2; carbohydrate processing enzyme of the endoplasmic reticulum; glucosidase II alpha-subunit; Glucosidase II subunit alpha; Neutral alpha-glucosidase AB
Gene Aliases: AU042638; G2AN; GANAB; GIIA; GLUII; KIAA0088; mKIAA0088
UniProt ID: (Human) Q14697, (Mouse) Q8BHN3
Entrez Gene ID: (Human) 23193, (Mouse) 14376
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support