Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEFPGLGSL GTSEPLPQFV DPALVSSTPE SGVFFPSGPE GLDAAASSTA PSTATAAAAA LAYYRDAEAY RHSPVFQVYP LLNCMEGIPG GSPYAGWAYG KTGLYPASTV CPTREDSPPQ AVEDLDGKGS TSFLETLKTE RLSPDLLTLG PALPSSLPVP NSAYGGPDFS STFFSPTGSP LNSAAYSSPK LRGTLPLPPC EARECVNCGA TATPLWRRDR TGHYLCNACG LYHKMNGQNR PLIRPKKRLI VSKRAGTQCT NCQTTTTTLW RRNASGDPVC NACGLYYKLH HQHYCGGSAQ LMRAQSMASR GGVVSFS (1-316 aa encoded by BC009797)
Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA 1 protein; GATA binding protein 1 (globin transcription factor 1); GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF-1; globin transcription factor 1; HGNC:4170; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1
Gene Aliases: ERYF1; GATA-1; GATA1; GF-1; GF1; NF-E1; NFE1; XLANP; XLTDA; XLTT
UniProt ID: (Human) P15976, (Mouse) P17679, (Rat) P43429
Entrez Gene ID: (Pig) 100620659, (Human) 2623, (Mouse) 14460, (Rat) 25172
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support