Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MATPNNLTPT NCSWWPISAL ESDAAKPAEA PDAPEAASPA HWPRESLVLY HWTQSFSSQK VRLVIAEKGL VCEERDVSLP QSEHKEPWFM RLNLGEEVPV IIHRDNIISD YDQIIDYVER TFTGEHVVAL MPEVGSLQHA RVLQYRELLD ALPMDAYTHG CILHPELTTD SMIPKYATAE IRRHLANATT DLMKLDHEEE PQLSEPYLSK QKKLMAKILE HDDVSYLKKI LGELAMVLDQ IEAELEKRKL ENEGQKCELW LCGCAFTLAD VLLGATLHRL KFLGLSKKYW EDGSRPNLQS FFERVQRRFA FRKVLGDIHT TLLSAVIPNA FRLVKRKPPS FFGASFLMGS LGGMGYFAYW YLKKKYI
The ganglioside GD3 synthase causes cell differentiation with neurite sprouting when transfected into the mouse neuroblastoma cell line Neuro2a. After differentiation, the expression of several genes is upregulated, including one that encodes a protein termed ganglioside-induced differentiation-associated protein 1 (Gdap1). A similar gene was found in humans, and mutations in the human gene are associated with Charcot-Marie-Tooth type 4A disease. The protein encoded by this gene is similar in sequence to the human GDAP1 protein. Several transcript variants encoding different isoforms, as well as a noncoding transcript variant, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ganglioside-induced differentiation-associated protein 1-like 1; GDAP1-L1; GDAP1L1; GIDAP1; GIDAP2; unnamed protein product
Gene Aliases: dJ881L22.1; dJ995J12.1.1; Gdap1l; GDAP1L1
UniProt ID: (Human) Q96MZ0
Entrez Gene ID: (Human) 78997, (Pig) 100625236, (Rabbit) 100347940, (Mouse) 228858, (Rat) 311616
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support