Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: DPLPTESRLM NSCLQARRKC QADPTCSAAY HHLDSCTSSI STPLPSEEPS VPADCLEAAQ QLRNSSLIGC MCHRRMKNQV ACLDIYWTVH RARSLGNYEL DVSPYEDTVT SKPWKMNLSK LNMLKPDSDL CLKFAMLCTL NDKCDRLRKA YGEACSGPHC QRHVCLRQLL TFFEKAAEPH AQGLLLCPCA PNDRGCGERR RNTIAPNCA
GFRa-3 binds persephin, thus, persephin, GFRa-3, and Ret PTK form a complex to transduce persephin signal and to mediate persephin function.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDNF family receptor alpha-3; GDNF receptor alpha-3; GDNF receptor alpha-3 variant; GDNFR-alpha-3; GFR alpha-3; GFR-alpha-3; GFRalpha3; glial cell line-derived neurotrophic factor receptor alpha-3; GPI-linked receptor; unnamed protein product
Gene Aliases: GDNFR3; GFRA3; GFRalpha3; UNQ339/PRO538/PRO3664; Y15110
UniProt ID: (Human) O60609, (Mouse) O35118
Entrez Gene ID: (Human) 2676, (Mouse) 14587
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support