Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Acts as a high affinity receptor for both nicotinic acid (also known as niacin) and (D)-beta-hydroxybutyrate and mediates increased adiponectin secretion and decreased lipolysis through G(i)-protein-mediated inhibition of adenylyl cyclase. This pharmacological effect requires nicotinic acid doses that are much higher than those provided by a normal diet. Mediates nicotinic acid-induced apoptosis in mature neutrophils. Receptor activation by nicotinic acid results in reduced cAMP levels which may affect activity of cAMP-dependent protein kinase A and phosphorylation of target proteins, leading to neutrophil apoptosis. The rank order of potency for the displacement of nicotinic acid binding is 5-methyl pyrazole-3-carboxylic acid = pyridine-3-acetic acid > acifran > 5-methyl nicotinic acid = acipimox >> nicotinuric acid = nicotinamide.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor HM74a; G-protein coupled receptor 109A; G-protein coupled receptor 109B; G-protein coupled receptor HM74; G-protein coupled receptor HM74A; G-protein coupled receptor HM74B; GTP-binding protein; hydroxy-carboxylic acid receptor 2; hydroxy-carboxylic acid receptor 3; niacin receptor 1; niacin receptor 2; nicotinic acid receptor; nicotinic acid receptor 2; putative chemokine receptor; unnamed protein product
Gene Aliases: GPR109A; GPR109B; HCA2; HCA3; HM74; HM74a; HM74b; NIACR1; Puma-g; PUMAG
UniProt ID: (Human) Q8TDS4, (Human) P49019
Entrez Gene ID: (Human) 338442, (Human) 8843
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support