PA5-29297 targets GRK6 in WB applications and shows reactivity with Human samples.
The PA5-29297 immunogen is recombinant fragment corresponding to a region within amino acids 335 and 556 of Human GRK6.
GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation. Several transcript variants encoding different isoforms have been described for GRK6.