Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MGLELFLDL VSQPSRAVYI FAKKNGIPLE LRTVDLVKGQ HKSKEFLQIN SLGKLPTLKD GDFILTESSA ILIYLSCKYQ TPDHWYPSDL QARARVHEYL GWHADCIRGT FGIPLWVQVL GPLIGVQVPE EKVERNRTAM DQALQWLEDK FLGDRPFLAG QQVTLADLMA LEELMQPVAL GYELFEGRPR LAAWRGRVEA FLGAELCQEA HSIILSILEQ AAKKTLPTPS PEAYQAMLLR IARIP (1-244 aa encoded by BC002415)
Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. Has a sulfatase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Em:AP000351.3; glutathione S-transferase 12; glutathione S-transferase theta-2-like; glutathione S-transferase Yrs-Yrs; GST 12-12; GST class-theta-2; GSTT2; unnamed protein product
Gene Aliases: mGSTT2; Yrs
UniProt ID: (Human) P0CG29, (Mouse) Q61133, (Rat) P30713
Entrez Gene ID: (Human) 2953, (Mouse) 14872, (Rat) 29487
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support