Product References
ERK and USP5 govern PD-1 homeostasis via deubiquitination to modulate tumor immunotherapy.
Nature communications
Xiao X,Shi J,He C,Bu X,Sun Y,Gao M,Xiang B,Xiong W,Dai P,Mao Q,Xing X,Yao Y,Yu H,Xu G,Li S,Ren Y,Chen B,Jiang C,Meng G,Lee YR,Wei W,Freeman GJ,Xie C,Zhang J
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Fri May 19 00:00:00 EDT 2023
Small-molecule PTPN2 Inhibitors Sensitize Resistant Melanoma to Anti-PD-1 Immunotherapy.
Cancer research communications
Zhu Z,Tang R,Huff S,Kummetha IR,Wang L,Li N,Rana TM
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2023
Combined PARP and WEE1 inhibition triggers anti-tumor immune response in BRCA1/2 wildtype triple-negative breast cancer.
NPJ breast cancer
Teo ZL,O'Connor MJ,Versaci S,Clarke KA,Brown ER,Percy LW,Kuykhoven K,Mintoff CP,Savas P,Virassamy B,Luen SJ,Byrne A,Sant S,Lindeman GJ,Darcy PK,Loi S
25-8898-82 was used in Flow cytometry/Cell sorting to identify baseline tumour-infiltrating lymphocyte (TIL) levels as a potential predictive biomarker of response to PARPi, WEE1i and immunotherapies in BRCA1/2 wild-type TNBC.
Tue Aug 15 00:00:00 EDT 2023
Meningeal lymphatics regulate radiotherapy efficacy through modulating anti-tumor immunity.
Cell research
Zhou C,Ma L,Xu H,Huo Y,Luo J
25-8898-82 was used in Flow Cytometry to suggest an essential role of the MLV-CLN network in RT-triggered anti-tumor immunity, and highlight the potential of VEGF-C mRNA for brain tumor therapy.
Wed Jun 01 00:00:00 EDT 2022
Histone methyltransferase WHSC1 loss dampens MHC-I antigen presentation pathway to impair IFN-γ-stimulated antitumor immunity.
The Journal of clinical investigation
Ren J,Li N,Pei S,Lian Y,Li L,Peng Y,Liu Q,Guo J,Wang X,Han Y,Zhang G,Wang H,Li Y,Jiang J,Li Q,Tan M,Peng J,Hu G,Xiao Y,Li X,Lin M,Qin J
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Fri Apr 15 00:00:00 EDT 2022
Wnt5A Signaling Blocks Progression of Experimental Visceral Leishmaniasis.
Frontiers in immunology
Maity S,Chakraborty A,Mahata SK,Roy S,Das AK,Sen M
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Mar 22 00:00:00 EDT 2022
Intranasal Delivery of Recombinant S100A8 Protein Delays Lung Cancer Growth by Remodeling the Lung Immune Microenvironment.
Frontiers in immunology
Wong SW,McCarroll J,Hsu K,Geczy CL,Tedla N
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Mon Jun 06 00:00:00 EDT 2022
eEF2K promotes PD-L1 stabilization through inactivating GSK3β in melanoma.
Journal for immunotherapy of cancer
Chen X,Wang K,Jiang S,Sun H,Che X,Zhang M,He J,Wen Y,Liao M,Li X,Zhou X,Song J,Ren X,Yi W,Yang J,Chen X,Yin M,Cheng Y
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2022
RAD21 amplification epigenetically suppresses interferon signaling to promote immune evasion in ovarian cancer.
The Journal of clinical investigation
Deng P,Wang Z,Chen J,Liu S,Yao X,Liu S,Liu L,Yu Z,Huang Y,Xiong Z,Xiao R,Gao J,Liang W,Chen J,Liu H,Hong JH,Chan JY,Guan P,Chen J,Wang Y,Yin J,Li J,Zheng M,Zhang C,Zhou P,Kang T,Teh BT,Yu Q,Zuo Z,Jiang Q,Liu J,Xiong Y,Xia X,Tan J
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Nov 15 00:00:00 EST 2022
PRDM1/BLIMP1 induces cancer immune evasion by modulating the USP22-SPI1-PD-L1 axis in hepatocellular carcinoma cells.
Nature communications
Li Q,Zhang L,You W,Xu J,Dai J,Hua D,Zhang R,Yao F,Zhou S,Huang W,Dai Y,Zhang Y,Baheti T,Qian X,Pu L,Xu J,Xia Y,Zhang C,Tang J,Wang X
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Mon Dec 12 00:00:00 EST 2022
Mobilizing phospholipids on tumor plasma membrane implicates phosphatidylserine externalization blockade for cancer immunotherapy.
Cell reports
Wang W,Wu S,Cen Z,Zhang Y,Chen Y,Huang Y,Cillo AR,Prokopec JS,Quarato G,Vignali DAA,Stewart-Ornstein J,Li S,Lu B,Gong YN
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
Enteric viruses evoke broad host immune responses resembling those elicited by the bacterial microbiome.
Cell host & microbe
Dallari S,Heaney T,Rosas-Villegas A,Neil JA,Wong SY,Brown JJ,Urbanek K,Herrmann C,Depledge DP,Dermody TS,Cadwell K
25-8898-82 was used in Flow cytometry/Cell sorting to expand an understanding of the immune space occupied by the enteric virome and underscore the importance of viral exposure events.
Wed Jun 09 00:00:00 EDT 2021
Blockade of 6-phosphogluconate dehydrogenase generates CD8+ effector T cells with enhanced anti-tumor function.
Cell reports
Daneshmandi S,Cassel T,Lin P,Higashi RM,Wulf GM,Boussiotis VA,Fan TW,Seth P
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Mar 09 00:00:00 EST 2021
Integrin αvβ8 on T cells suppresses anti-tumor immunity in multiple models and is a promising target for tumor immunotherapy.
Cell reports
Dodagatta-Marri E,Ma HY,Liang B,Li J,Meyer DS,Chen SY,Sun KH,Ren X,Zivak B,Rosenblum MD,Headley MB,Pinzas L,Reed NI,Del Cid JS,Hann BC,Yang S,Giddabasappa A,Noorbehesht K,Yang B,Dal Porto J,Tsukui T,Niessen K,Atakilit A,Akhurst RJ,Sheppard D
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Jul 06 00:00:00 EDT 2021
Aiduqing formula inhibits breast cancer metastasis by suppressing TAM/CXCL1-induced Treg differentiation and infiltration.
Cell communication and signaling : CCS
Li J,Wang S,Wang N,Zheng Y,Yang B,Wang X,Zhang J,Pan B,Wang Z
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Mon Aug 30 00:00:00 EDT 2021
Extracellular Vesicles from Akkermansia muciniphila Elicit Antitumor Immunity Against Prostate Cancer via Modulation of CD8+ T Cells and Macrophages.
International journal of nanomedicine
Luo ZW,Xia K,Liu YW,Liu JH,Rao SS,Hu XK,Chen CY,Xu R,Wang ZX,Xie H
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Non-redundant activity of GSK-3α and GSK-3β in T cell-mediated tumor rejection.
iScience
Steele L,Mannion AJ,Shaw G,Maclennan KA,Cook GP,Rudd CE,Taylor A
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Fri Jun 25 00:00:00 EDT 2021
Non-redundant activity of GSK-3α and GSK-3β in T cell-mediated tumor rejection.
iScience
Steele L,Mannion AJ,Shaw G,Maclennan KA,Cook GP,Rudd CE,Taylor A
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Fri Jun 25 00:00:00 EDT 2021
Circadian Clock Disruption Suppresses PDL1+ Intraepithelial B Cells in Experimental Colitis and Colitis-Associated Colorectal Cancer.
Cellular and molecular gastroenterology and hepatology
Liu JL,Wang CY,Cheng TY,Rixiati Y,Ji C,Deng M,Yao S,Yuan LH,Zhao YY,Shen T,Li JM
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Mon Jan 24 00:00:00 EST 2022
Knockout of immunotherapy prognostic marker genes eliminates the effect of the anti-PD-1 treatment.
NPJ precision oncology
Yang N,Ji F,Cheng L,Lu J,Sun X,Lin X,Lan X
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Fri May 07 00:00:00 EDT 2021
Metabolic requirements of NK cells during the acute response against retroviral infection.
Nature communications
Littwitz-Salomon E,Moreira D,Frost JN,Choi C,Liou KT,Ahern DK,O'Shaughnessy S,Wagner B,Biron CA,Drakesmith H,Dittmer U,Finlay DK
25-8898-82 was used in Flow cytometry/Cell sorting to show that following infection natural killer cells increase nutrient uptake, including amino acids and iron, and reprogram their metabolic machinery by increasing glycolysis and mitochondrial metabolism.
Fri Sep 10 00:00:00 EDT 2021
Assessment of the WAP-Myc mouse mammary tumor model for spontaneous metastasis.
Scientific reports
Utz B,Turpin R,Lampe J,Pouwels J,Klefström J
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Fri Oct 30 00:00:00 EDT 2020
Cancer-Specific Loss of p53 Leads to a Modulation of Myeloid and T Cell Responses.
Cell reports
Blagih J,Zani F,Chakravarty P,Hennequart M,Pilley S,Hobor S,Hock AK,Walton JB,Morton JP,Gronroos E,Mason S,Yang M,McNeish I,Swanton C,Blyth K,Vousden KH
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Jan 14 00:00:00 EST 2020
Cancer-Specific Loss of p53 Leads to a Modulation of Myeloid and T Cell Responses.
Cell reports
Blagih J,Zani F,Chakravarty P,Hennequart M,Pilley S,Hobor S,Hock AK,Walton JB,Morton JP,Gronroos E,Mason S,Yang M,McNeish I,Swanton C,Blyth K,Vousden KH
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Jan 14 00:00:00 EST 2020
Salmonella-Based Therapy Targeting Indoleamine 2,3-Dioxygenase Restructures the Immune Contexture to Improve Checkpoint Blockade Efficacy.
Biomedicines
Ebelt ND,Zuniga E,Marzagalli M,Zamloot V,Blazar BR,Salgia R,Manuel ER
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Wed Dec 16 00:00:00 EST 2020
Memory CD73+IgM+ B cells protect against Plasmodium yoelii infection and express Granzyme B.
PloS one
Parra M,Weitner M,Yang A,Akue A,Liu X,Schmidt T,Allman WR,Akkoyunlu M,Derrick SC
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Oct 27 00:00:00 EDT 2020
GAPDH Overexpression in the T Cell Lineage Promotes Angioimmunoblastic T Cell Lymphoma through an NF-κB-Dependent Mechanism.
Cancer cell
Mondragón L,Mhaidly R,De Donatis GM,Tosolini M,Dao P,Martin AR,Pons C,Chiche J,Jacquin M,Imbert V,Proïcs E,Boyer L,Doye A,Luciano F,Neels JG,Coutant F,Fabien N,Sormani L,Rubio-Patiño C,Bossowski JP,Muller F,Marchetti S,Villa E,Peyron JF,Gaulard P,Lemonnier F,Asnafi V,Genestier L,Benhida R,Fournié JJ,Passeron T,Ricci JE,Verhoeyen E
25-8898-82 was used in Flow Cytometry to suggest the therapeutic potential of targeting NF-κB signaling in AITL and provide a model for future AITL therapeutic investigations.
Mon Sep 16 00:00:00 EDT 2019
Inflammation induced by incomplete radiofrequency ablation accelerates tumor progression and hinders PD-1 immunotherapy.
Nature communications
Shi L,Wang J,Ding N,Zhang Y,Zhu Y,Dong S,Wang X,Peng C,Zhou C,Zhou L,Li X,Shi H,Wu W,Long X,Wu C,Liao W
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Thu Nov 28 00:00:00 EST 2019
Qa-1-Restricted CD8+ T Cells Can Compensate for the Absence of Conventional T Cells during Viral Infection.
Cell reports
Anderson CK,Reilly EC,Lee AY,Brossay L
25-8898 was used in Flow cytometry/Cell sorting to study compensatory mechanisms in immune responses to viruses when classical T cell immunity has been evaded.
Tue Apr 09 00:00:00 EDT 2019
Pharmacological reactivation of MYC-dependent apoptosis induces susceptibility to anti-PD-1 immunotherapy.
Nature communications
Haikala HM,Anttila JM,Marques E,Raatikainen T,Ilander M,Hakanen H,Ala-Hongisto H,Savelius M,Balboa D,Von Eyss B,Eskelinen V,Munne P,Nieminen AI,Otonkoski T,Schüler J,Laajala TD,Aittokallio T,Sihto H,Mattson J,Heikkilä P,Leidenius M,Joensuu H,Mustjoki S,Kovanen P,Eilers M,Leverson JD,Klefström J
25-8898-82 was used in Flow Cytometry to demonstrate that pharmacological reactivation of MYC-dependent apoptosis is a powerful antitumor strategy involving both tumor cell depletion and immunosurveillance.
Wed Feb 06 00:00:00 EST 2019
The Transcription Factor Runx3 Establishes Chromatin Accessibility of cis-Regulatory Landscapes that Drive Memory Cytotoxic T Lymphocyte Formation.
Immunity
Wang D,Diao H,Getzler AJ,Rogal W,Frederick MA,Milner J,Yu B,Crotty S,Goldrath AW,Pipkin ME
25-8898 was used in Flow cytometry/Cell sorting to conclude that Runx3 governs chromatin accessibility during TCR stimulation and enforces the memory CTL developmental program.
Tue Apr 17 00:00:00 EDT 2018
MAIT cells contribute to protection against lethal influenza infection in vivo.
Nature communications
van Wilgenburg B,Loh L,Chen Z,Pediongco TJ,Wang H,Shi M,Zhao Z,Koutsakos M,Nüssing S,Sant S,Wang Z,D'Souza C,Jia X,Almeida CF,Kostenko L,Eckle SBG,Meehan BS,Kallies A,Godfrey DI,Reading PC,Corbett AJ,McCluskey J,Klenerman P,Kedzierska K,Hinks TSC
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Fri Nov 09 00:00:00 EST 2018
Crosstalks between mTORC1 and mTORC2 variagate cytokine signaling to control NK maturation and effector function.
Nature communications
Wang F,Meng M,Mo B,Yang Y,Ji Y,Huang P,Lai W,Pan X,You T,Luo H,Guan X,Deng Y,Yuan S,Chu J,Namaka M,Hughes T,Ye L,Yu J,Li X,Deng Y
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Mon Nov 19 00:00:00 EST 2018
Regulation of chitinase-3-like-1 in T cell elicits Th1 and cytotoxic responses to inhibit lung metastasis.
Nature communications
Kim DH,Park HJ,Lim S,Koo JH,Lee HG,Choi JO,Oh JH,Ha SJ,Kang MJ,Lee CM,Lee CG,Elias JA,Choi JM
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Mon Feb 05 00:00:00 EST 2018
Inflammatory Ly6Chigh Monocytes Protect against Candidiasis through IL-15-Driven NK Cell/Neutrophil Activation.
Immunity
Domínguez-Andrés J,Feo-Lucas L,Minguito de la Escalera M,González L,López-Bravo M,Ardavín C
25-8898 was used in Flow cytometry/Cell sorting to explore the role of inflammatory Ly6Chigh monocytes in natural killer cell-mediated neutrophil activation during the innate immune response against Candida albicans.
Tue Jun 20 00:00:00 EDT 2017
Combined antiangiogenic and anti-PD-L1 therapy stimulates tumor immunity through HEV formation.
Science translational medicine
Allen E,Jabouille A,Rivera LB,Lodewijckx I,Missiaen R,Steri V,Feyen K,Tawney J,Hanahan D,Michael IP,Bergers G
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Wed Apr 12 00:00:00 EDT 2017
Egr2 and 3 control adaptive immune responses by temporally uncoupling expansion from T cell differentiation.
The Journal of experimental medicine
Miao T,Symonds ALJ,Singh R,Symonds JD,Ogbe A,Omodho B,Zhu B,Li S,Wang P
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Mon Jun 05 00:00:00 EDT 2017
1810011o10 Rik Inhibits the Antitumor Effect of Intratumoral CD8+ T Cells through Suppression of Notch2 Pathway in a Murine Hepatocellular Carcinoma Model.
Frontiers in immunology
Dai K,Huang L,Huang YB,Chen ZB,Yang LH,Jiang YA
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
Targeted calcium influx boosts cytotoxic T lymphocyte function in the tumour microenvironment.
Nature communications
Kim KD,Bae S,Capece T,Nedelkovska H,de Rubio RG,Smrcka AV,Jun CD,Jung W,Park B,Kim TI,Kim M
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Mon May 15 00:00:00 EDT 2017
Loss of HIF-1α in natural killer cells inhibits tumour growth by stimulating non-productive angiogenesis.
Nature communications
Krzywinska E,Kantari-Mimoun C,Kerdiles Y,Sobecki M,Isagawa T,Gotthardt D,Castells M,Haubold J,Millien C,Viel T,Tavitian B,Takeda N,Fandrey J,Vivier E,Sexl V,Stockmann C
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Fri Nov 17 00:00:00 EST 2017
Lack of the programmed death-1 receptor renders host susceptible to enteric microbial infection through impairing the production of the mucosal natural killer cell effector molecules.
Journal of leukocyte biology
Solaymani-Mohammadi S,Lakhdari O,Minev I,Shenouda S,Frey BF,Billeskov R,Singer SM,Berzofsky JA,Eckmann L,Kagnoff MF
25-8898 was used in Flow cytometry/Cell sorting to identify the role of the programmed death-1 receptor in early host microbial defence at the intestinal mucosa.
Tue Mar 01 00:00:00 EST 2016
The stress kinase GCN2 does not mediate suppression of antitumor T cell responses by tryptophan catabolism in experimental melanomas.
Oncoimmunology
Sonner JK,Deumelandt K,Ott M,Thomé CM,Rauschenbach KJ,Schulz S,Munteanu B,Mohapatra S,Adam I,Hofer AC,Feuerer M,Opitz CA,Hopf C,Wick W,Platten M
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Caribbean and La Réunion Chikungunya Virus Isolates Differ in Their Capacity To Induce Proinflammatory Th1 and NK Cell Responses and Acute Joint Pathology.
Journal of virology
Teo TH,Her Z,Tan JJ,Lum FM,Lee WW,Chan YH,Ong RY,Kam YW,Leparc-Goffart I,Gallian P,Rénia L,de Lamballerie X,Ng LF
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
Intestinal microbiota modulates gluten-induced immunopathology in humanized mice.
The American journal of pathology
Galipeau HJ,McCarville JL,Huebener S,Litwin O,Meisel M,Jabri B,Sanz Y,Murray JA,Jordana M,Alaedini A,Chirdo FG,Verdu EF
25-8898 was used in Flow cytometry/Cell sorting to assess whether specific microbiota compositions influence immune responses to gluten in mice expressing the human DQ8 gene, which confers moderate CD genetic susceptibility.
Sun Nov 01 00:00:00 EDT 2015
mTORC1-dependent metabolic reprogramming is a prerequisite for NK cell effector function.
Journal of immunology (Baltimore, Md. : 1950)
Donnelly RP,Loftus RM,Keating SE,Liou KT,Biron CA,Gardiner CM,Finlay DK
25-8898 was used in Flow cytometry/Cell sorting to investigate whether mTORC1-controlled metabolism underpins normal natural killer cell proinflammatory functions.
Sat Nov 01 00:00:00 EDT 2014
Damage to the blood-brain barrier during experimental cerebral malaria results from synergistic effects of CD8+ T cells with different specificities.
Infection and immunity
Poh CM,Howland SW,Grotenbreg GM,Rénia L
25-8898 was used in Immunocytochemistry to show that blood-brain barrier damage during experimental cerebral malaria is mediated by synergistic actions of CD8T cells recognizing different malaria-specific epitopes.
Sat Nov 01 00:00:00 EDT 2014
Metastasis is regulated via microRNA-200/ZEB1 axis control of tumour cell PD-L1 expression and intratumoral immunosuppression.
Nature communications
Chen L,Gibbons DL,Goswami S,Cortez MA,Ahn YH,Byers LA,Zhang X,Yi X,Dwyer D,Lin W,Diao L,Wang J,Roybal J,Patel M,Ungewiss C,Peng D,Antonia S,Mediavilla-Varela M,Robertson G,Suraokar M,Welsh JW,Erez B,Wistuba II,Chen L,Peng D,Wang S,Ullrich SE,Heymach JV,Kurie JM,Qin FX
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Tue Oct 28 00:00:00 EDT 2014
Tim-3 negatively mediates natural killer cell function in LPS-induced endotoxic shock.
PloS one
Hou H,Liu W,Wu S,Lu Y,Peng J,Zhu Y,Lu Y,Wang F,Sun Z
Published figure using Granzyme B monoclonal antibody (Product # 25-8898-82) in Flow Cytometry
Mon Jul 06 00:00:00 EDT 2015
Both NK cell-intrinsic and -extrinsic STAT1 signaling are required for NK cell response against vaccinia virus.
Journal of immunology (Baltimore, Md. : 1950)
Fortin C,Huang X,Yang Y
25-8898 was used in Flow cytometry/Cell sorting to explore the role of STAT1 in natural killer cell responses to vaccinia virus infection in vivo.
Mon Jul 01 00:00:00 EDT 2013