Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Growth Hormone Receptor Antibodies

          Invitrogen

          Growth Hormone Receptor Polyclonal Antibody

          View all (25) Growth Hormone Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Growth Hormone Receptor Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Growth Hormone Receptor Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Growth Hormone Receptor Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Growth Hormone Receptor Antibody (PA5-79309) in WB

          Western blot analysis of Growth Hormone Receptor in Lane 1: rat liver tissue lysate, Lane 2: rat kidney tissue lysate, Lane 3: rat spleen tissue lysate, Lane 4: rat Intestine tissue lysate, Lane 5: mouse spleen tissue lysate, Lane 6: mouse testis tissue lysate, Lane 7: mouse liver tissue lysate, Lane 8: mouse kidney tissue lysate, Lane 9: mouse Intestine tissue lysate using 50 µg per well. Sample was incuba... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Growth Hormone Receptor Antibody in Western Blot (WB)
          Growth Hormone Receptor Polyclonal Antibody

          Product Details

          PA5-79309

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide corresponding to a sequence in the middle region of mouse Growth hormone receptor (231-247aa DKEHEVRVRSRQRSFEK).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5mg BSA

          Contains

          0.05mg sodium azide, 0.05mg thimerosal

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2746425

          Product Specific Information

          Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

          Positive Control - WB: Rat Liver Tissue, Rat Kidney Tissue, Rat Spleen Tissue, Rat Intestine Tissue, Mouse Spleen Tissue, Mouse Testis Tissue, Mouse Liver Tissue, Mouse Kidney Tissue, Mouse Intestine Tissue.

          Target Information

          Growth hormone receptor (GHR) is a gene located on chromosome 5p13.1, encoding a transmembrane receptor that plays a critical role in mediating the effects of growth hormone, which is crucial for growth, metabolism, and tissue development. GHR is expressed in various tissues, including the liver, muscle, and adipose tissue, and facilitates the growth hormone signaling pathway by binding to growth hormone, leading to receptor dimerization and activation of intracellular signaling cascades such as the JAK2-STAT5 pathway. This signaling promotes growth and anabolic processes, influencing body composition, insulin sensitivity, and lipid metabolism. Mutations or alterations in the GHR gene can lead to growth disorders like Laron syndrome, characterized by insensitivity to growth hormone. Understanding GHR's structure and function is pivotal for developing therapeutic strategies for growth deficiencies and metabolic syndromes, as modifying GHR signaling pathways can offer potential interventions in related conditions.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GH receptor; Growth hormone receptor; Growth hormone receptor precursor (GH receptor) (GH binding protein) (GHBP) (Serum binding protein); growth hormone receptor precursor splice variant D56; growth hormone receptor variant d5-6; growth hormone receptor/binding protein; MGC124963; MGC156665; Somatotropin receptor

          View more View less

          Gene Aliases: GHBP; Ghr; GHR/BP

          View more View less

          UniProt ID: (Rat) P16310, (Mouse) P16882

          View more View less

          Entrez Gene ID: (Rat) 25235, (Mouse) 14600

          View more View less

          Function(s)
          growth hormone receptor activity peptide hormone binding growth factor binding protein kinase binding protein phosphatase binding SH2 domain binding protein homodimerization activity cytokine receptor activity protein binding transmembrane signal receptor
          Process(es)
          activation of MAPK activity allantoin metabolic process citrate metabolic process 2-oxoglutarate metabolic process succinate metabolic process oxaloacetate metabolic process isoleucine metabolic process valine metabolic process creatine metabolic process fatty acid metabolic process JAK-STAT cascade response to hormone hormone-mediated signaling pathway cytokine-mediated signaling pathway taurine metabolic process receptor internalization response to food response to estradiol cellular response to insulin stimulus cellular response to hormone stimulus response to cytokine regulation of multicellular organism growth positive regulation of multicellular organism growth activation of Janus kinase activity activation of JAK2 kinase activity response to morphine response to peptide hormone multicellular organism metabolic process positive regulation of cell differentiation positive regulation of JAK-STAT cascade creatinine metabolic process response to cycloheximide insulin-like growth factor receptor signaling pathway positive regulation of peptidyl-tyrosine phosphorylation response to glucocorticoid cartilage development involved in endochondral bone morphogenesis growth hormone receptor signaling pathway response to interleukin-1 negative regulation of neuron death endocytosis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0