Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HCN1 Antibodies

          Osenses

          HCN1 Polyclonal Antibody

          2 References
          View all (25) HCN1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite HCN1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          OSH00011W-100UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-1:2,000
          -

          Immunohistochemistry (IHC)

          1:300-1:2,000
          -

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Sheep / Ig

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from cytoplasmic region of human HCN1 conjugated to an immunogenic carrier protein was used as the immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

          The antigen is homologous in many species including mouse and rat.

          Specificity of this antibody: HCN1.

          Target Information

          HCN1 includes Hyperpolarization-activated cation channels of the HCN gene family, which contribute to spontaneous rhythmic activity in both heart and brain. HCN1 is a member of a family of pacemaker channels activated by hyperpolarization and regulated by cyclic nucleotides. HCN1 and HCN2 play an important role for motor learning and neuronal integration by cerebellar Purkinje cells as well as, shaping autonomous activity of single neurons and the periodicity of network oscillations. The HCN1 expression is highly enriched in cerebral cortex, hippocampus, cerebellum, and facial motor nucleus. HCN2 is highly abundant in mamillary bodies, pontine nucleus, ventral cochlear nucleus, and nucleus of the trapezoid body. These variations in regional specificity of HCN channels could generate important differences in neuronal pacemaker activity across brain systems.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: BCNG-1; Brain cyclic nucleotide-gated channel 1; HAC-2; Hyperpolarization-activated cation channel 2; potassium channel; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1

          View more View less

          Gene Aliases: BCNG-1; BCNG1; C630013B14Rik; DEE24; EIEE24; GEFSP10; HAC-2; HAC2; HCN1

          View more View less

          UniProt ID: (Human) O60741, (Mouse) O88704, (Rat) Q9JKB0

          View more View less

          Entrez Gene ID: (Human) 348980, (Mouse) 15165, (Rat) 84390

          View more View less

          Function(s)
          nucleotide binding ion channel activity intracellular cAMP activated cation channel activity voltage-gated sodium channel activity voltage-gated potassium channel activity potassium channel activity sodium channel activity protein binding phosphatidylinositol-4,5-bisphosphate binding phosphatidylinositol-3,4,5-trisphosphate binding voltage-gated cation channel activity cAMP binding identical protein binding intracellular cAMP-activated cation channel activity involved in regulation of presynaptic membrane potential voltage-gated ion channel activity protein C-terminus binding protein complex binding ion channel transporter
          Process(es)
          regulation of membrane depolarization ion transport potassium ion transport sodium ion transport neuronal action potential ion transmembrane transport cellular response to interferon-beta sodium ion transmembrane transport regulation of membrane potential maternal behavior apical protein localization negative regulation of action potential retinal cone cell development protein homotetramerization response to calcium ion general adaptation syndrome, behavioral process negative regulation of synaptic transmission, glutamatergic transmembrane transport regulation of postsynaptic membrane potential cellular response to cAMP potassium ion transmembrane transport regulation of heart rate by cardiac conduction sodium ion import across plasma membrane regulation of SA node cell action potential response to L-glutamate regulation of membrane hyperpolarization positive regulation of membrane hyperpolarization potassium ion import across plasma membrane transport regulation of ion transmembrane transport positive regulation of cation channel activity inflammatory response aging response to cisplatin
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline