Hepatitis E virus capsid is made up of individual subunits consisting of homodimers of a single structural protein forming the virus shell. These dimers are believed to protrude from the viral surface and to interact with host cells to initiate infection.