    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD154 (CD40 Ligand) Antibodies
          Invitrogen
          CD154 (CD40 Ligand) Monoclonal Antibody (MR1), FITC
          2 References
          View all (105) CD154 (CD40 Ligand) antibodies
          Questions & Answers
          Cite CD154 (CD40 Ligand) Monoclonal Antibody (MR1), FITC
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          CD154 (CD40 Ligand) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD154 (CD40 Ligand) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CD154 (CD40 Ligand) Antibody (HMCD15401) in Flow
          Flow cytometry analysis of CD154 (CD40 Ligand) in PMA and ionomycin stimulated BALB/c mouse splenocytes. Cells were stained with CD154 (CD40 Ligand) Monoclonal Antibody (MR1), FITC (Product # HMCD15401), Rat Anti-Mouse CD4-PE , and Rat Anti-Mouse CD8 alpha-PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD154 (CD40 Ligand) Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          CD154 (CD40 Ligand) Monoclonal Antibody (MR1), FITC
          Product Details
          HMCD15401
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          -
          View 1 publication 1 publication
          Flow Cytometry (Flow)
          0.1-0.2 µg / 1x10^6 cells
          -
          ELISA (ELISA)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Armenian hamster / IgG3
          Class
          Monoclonal
          Type
          Antibody
          Clone
          MR1
          Immunogen
          Mouse CD154. from Activated mouse Th1 clone D1.6
          3D Epitope / Immunogen
          Conjugate
          FITC FITC FITC
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          498/517 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          500 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.4
          Contains
          <0.1% sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536595
          Target Information
          CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: H-CD-40-L; M-CD-40-L; membrane protein; RP23-153G22.3; T-cell antigen Gp39; TNF-related activation protein; tumor necrosis factor ligand 8b; tumor necrosis factor ligand superfamily member 5
          View more View less
          Gene Aliases: CD154; CD40-L; Cd40l; gp39; HIGM1; IGM; IMD3; Ly-62; Ly62; T-BAM; Tnfsf5; Tnlg8b; TRAP
          View more View less
          UniProt ID: (Mouse) P27548
          View more View less
          Entrez Gene ID: (Mouse) 21947
          View more View less
          Function(s)
          cytokine activity tumor necrosis factor receptor binding CD40 receptor binding integrin binding protein serine/threonine kinase activator activity
          Process(es)
          regulation of immunoglobulin production regulation of immune system process positive regulation of immune response to tumor cell inflammatory response immune response signal transduction cell surface receptor signaling pathway integrin-mediated signaling pathway regulation of gene expression positive regulation of gene expression CD40 signaling pathway platelet activation B cell differentiation positive regulation of B cell proliferation positive regulation of interferon-gamma production positive regulation of interleukin-10 production positive regulation of interleukin-12 production positive regulation of interleukin-4 production B cell proliferation positive regulation of T cell proliferation negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling isotype switching positive regulation of MHC class II biosynthetic process dendritic cell differentiation positive regulation of endothelial cell apoptotic process positive regulation of dendritic cell differentiation positive regulation of extrinsic apoptotic signaling pathway
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch