Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: TGGVGAVAL LIGPNAPLIF ERGLRGTHMQ HAYDFYKPDM LSEYPIVDGK LSIQCYLSAL DRCYSVYCKK IHAQWQKEGN DKDFTLNDFG FMIFHSPYCK LVQKSLARML LNDFLNDQNR DKNSIYSGLE AFGDVKLEDT YFDRDVEKAF MKASSELFSQ KTKASLLVSN QNGNMYTSSV YGSLASVLAQ YSPQQLAGKR IGVFSYGSGL AATLYSLKVT QDATPGSALD KITASLCDLK SRLDSRTGVA PDVFAENMKL REDTYHLVNY IPQGSIDSLF EGTWYLVRVD EKHRRTYARR PTPNDDTLDE GVGLVHSNIA TEHIPSPAKK VPRLPATAAE PEAAVISNGE H (171-520 aa encoded by BC083514)
This enzyme condenses acetyl-CoA with acetoacetyl-CoA to form HMG-CoA, which is the substrate for HMG-CoA reductase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble); HMG-CoA synthase; hydroxymethylglutaryl-CoA synthase 1; unnamed protein product
Gene Aliases: B130032C06Rik; CMYO28; HMGCS
UniProt ID: (Human) Q01581, (Rat) P17425, (Mouse) Q8JZK9
Entrez Gene ID: (Human) 3157, (Rat) 29637, (Mouse) 208715
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support