Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAAPALRLC HIAFHVPAGQ PLARNLQRLF GFQPLASREV DGWRQLALRS GDAVFLVNEG AGSGEPLYGL DPRHAVPSAT NLCFDVADAG AATRELAALG CSVPVPPVRV RDAQGAATYA VVSSPAGILS LTLLERAGYR GPFLPGFRPV SSAPGPGWVS RVDHLTLACT PGSSPTLLRW FHDCLGFCHL PLSPGEDPEL GLEMTAGFGL GGLRLTALQA QPGSIVPTLV LAESLPGATT RQDQVEQFLA RHKGPGLQHV GLYTPNIVEA TEGVATAGGQ FLAPPGAYYQ QPGKERQIRA AGHEPHLLAR QGILLDGDKG KFLLQVFTKS LFTEDTFFLE LIQRQGATGF GQGNIRALWQ SVQEQSARSQ EA (1-371 aa encoded by BC007293)
May have dioxygenase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glyoxalase domain-containing protein 1; HPD-like protein; RP4-534D1.1; unnamed protein product
Gene Aliases: 4-HPPD-L; GLOXD1; NEDSWMA; SPG83
UniProt ID: (Human) Q96IR7
Entrez Gene ID: (Human) 84842
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support