Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IFNAR1 Antibodies

          Abnova

          IFNAR1 Polyclonal Antibody, MaxPab™

          View all (46) IFNAR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IFNAR1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          IFNAR1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IFNAR1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IFNAR1 Antibody (H00003454-D01P) in WB

          Western Blot analysis of IFNAR1 expression in transfected 293T cell line by IFNAR1 MaxPab polyclonal antibody. Lane 1: IFNAR1 transfected lysate(63.50 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IFNAR1 Antibody in Western Blot (WB)
          IFNAR1 Polyclonal Antibody, MaxPab™

          Product Details

          H00003454-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          IFNAR1 (NP_000620.2, 1 a.a. approximately 557 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MMVVLLGATT LVLVAVAPWV LSAAAGGKNL KSPQKVEVDI IDDNFILRWN RSDESVGNVT FSFDYQKTGM DNWIKLSGCQ NITSTKCNFS SLKLNVYEEI KLRIRAEKEN TSSWYEVDSF TPFRKAQIGP PEVHLEAEDK AIVIHISPGT KDSVMWALDG LSFTYSLVIW KNSSGVEERI ENIYSRHKIY KLSPETTYCL KVKAALLTSW KIGVYSPVHC IKTTVENELP PPENIEVSVQ NQNYVLKWDY TYANMTFQVQ WLHAFLKRNP GNHLYKWKQI PDCENVKTTQ CVFPQNVFQK GIYLLRVQAS DGNNTSFWSE EIKFDTEIQA FLLPPVFNIR SLSDSFHIYI GAPKQSGNTP VIQDYPLIYE IIFWENTSNA ERKIIEKKTD VTVPNLKPLT VYCVKARAHT MDEKLNKSSV FSDAVCEKTK PGNTSKIWLI VGICIALFAL PFVIYAAKVF LRCINYVFFP SLKPSSSIDE YFSEQPLKNL LLSTSEEQIE KCFIIENIST IATVEETNQT DEDHKKYSSQ TSQDSGNYSN EDESESKTSE ELQQDFV

          Target Information

          Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001, Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alpha-type antiviral protein; beta-type antiviral protein; CRF2-1; Cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; H3K1; IFN R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFN-R-1; IFNalpha/beta R1; IFNalpha/beta receptor 1; IFNalpha/beta-Ralpha; IFNalpha/betaR1; IFNR1; interferon (alpha, beta and omega) receptor 1; Interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha receptor precursor; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; Type I interferon receptor 1; unnamed protein product

          View more View less

          Gene Aliases: AVP; IFN-alpha-REC; IFNAR; IFNAR1; IFNBR; IFRC; IMD106

          View more View less

          UniProt ID: (Human) P17181

          View more View less

          Entrez Gene ID: (Human) 3454

          View more View less

          Function(s)
          interferon receptor activity type I interferon receptor activity protein binding JAK pathway signal transduction adaptor activity cytokine binding type I interferon binding
          Process(es)
          regulation of transcription from RNA polymerase II promoter JAK-STAT cascade response to virus positive regulation of metabolic process cytokine-mediated signaling pathway response to lipopolysaccharide cellular response to interferon-alpha cellular response to interferon-beta regulation of JAK-STAT cascade type I interferon signaling pathway cellular response to virus positive regulation of protein localization to nucleus positive regulation of cellular respiration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline