Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ITIH1 Antibodies

          Abnova

          ITIH1 Polyclonal Antibody, MaxPab™

          View all (7) ITIH1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ITIH1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ITIH1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ITIH1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ITIH1 Antibody (H00003697-B01P) in WB

          Western Blot analysis of ITIH1 expression in transfected 293T cell line by ITIH1 MaxPab polyclonal antibody. Lane1:ITIH1 transfected lysate(100.21 KDa). Lane2:Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ITIH1 Antibody in Western Blot (WB)

          Product Details

          H00003697-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ITIH1 (NP_002206.1, 1 a.a. approximately 911 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MDGAMGPRGL LLCMYLVSLL ILQAMPALGS ATGRSKSSEK RQAVDTAVDG VFIRSLKVNC KVTSRFAHYV VTSQVVNTAN EAREVAFDLE IPKTAFISDF AVTADGNAFI GDIKDKVTAW KQYRKAAISG ENAGLVRASG RTMEQFTIHL TVNPQSKVTF QLTYEEVLKR NHMQYEIVIK VKPKQLVHHF EIDVDIFEPQ GISKLDAQAS FLPKELAAQT IKKSFSGKKG HVLFRPTVSQ QQSCPTCSTS LLNGHFKVTY DVSRDKICDL LVANNHFAHF FAPQNLTNMN KNVVFVIDIS GSMRGQKVKQ TKEALLKILG DMQPGDYFDL VLFGTRVQSW KGSLVQASEA NLQAAQDFVR GFSLDEATNL NGGLLRGIEI LNQVQESLPE LSNHASILIM LTDGDPTEGV TDRSQILKNV RNAIRGRFPL YNLGFGHNVD FNFLEVMSME NNGRAQRIYE DHDATQQLQG FYSQVAKPLL VDVDLQYPQD AVLALTQNHH KQYYEGSEIV VAGRIADNKQ SSFKADVQAH GEGQEFSITC LVDEEEMKKL LRERGHMLEN HVERLWAYLT IQELLAKRMK VDREVRANLS SQALRMSLDY GFVTPLTSMS IRGMADQDGL KPTIDKPSED SPPLEMLGPR RTFVLSALQP SPTHSSSNTQ RLPDRVTGVD TDPHFIIHVP QKEDTLCFNI NEEPGVILSL VQDPNTGFSV NGQLIGNKAR SPGQHDGTYF GRLGIANPAT DFQLEVTPQN ITLNPGFGGP VFSWRDQAVL RQDGVVVTIN KKRNLVVSVD DGGTFEVVLH RVWKGSSVHQ DFLGFYVLDS HRMSARTHGL LGQFFHPIGF EVSDIHPGSD PTKPDATMVV RNRRLTVTRG LQKDYSKDPW HGAEVSCWFI HNNGAGLIDG AYTDYIVPDI F

          Target Information

          The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CYCSP39; HCP39; inter-alpha (globulin) inhibitor, H1 polypeptide; inter-alpha-inhibitor heavy chain 1; Inter-alpha-trypsin inhibitor complex component III; Inter-alpha-trypsin inhibitor heavy chain H1; inter-alpha-trypsin inhibitor heavy chain old gene name 'ITI'; ITI heavy chain H1; MGC126415; predicted protein of HQ2769; Serum-derived hyaluronan-associated protein; SHAP; unnamed protein product

          View more View less

          Gene Aliases: H1P; IATIH; IGHEP1; ITI-HC1; ITIH; ITIH1; SHAP

          View more View less

          UniProt ID: (Human) P19827

          View more View less

          Entrez Gene ID: (Human) 3697

          View more View less

          Function(s)
          serine-type endopeptidase inhibitor activity calcium ion binding protein binding hyaluronic acid binding carbohydrate binding peptidase inhibitor activity
          Process(es)
          hyaluronan metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline