Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta OL; beta oligodendroglia; CD49 antigen-like family member E; collagen alpha-1(I) chain-like; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor subunit beta; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; glycoprotein IIa; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-F; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta-1 subunit (cytoplasmic domain); Integrin beta1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITGA5/SLC9B1 fusion; surface glycoprotein receptor; unnamed protein product; very late activation protein 5, alpha subunit; very late activation protein, beta polypeptide; VLA-4 subunit beta
Gene Aliases: 4633401G24Rik; CD29; CD49e; FNRA; FNRB; Gm9863; GPIIA; Itga5_retired; MDF2; MSK12; VLA-5; VLA-BETA; VLA5; VLA5A; VLAB
UniProt ID: (Human) P08648, (Human) P05556, (Mouse) P11688, (Rat) P49134, (Mouse) P09055
Entrez Gene ID: (Human) 3678, (Human) 3688, (Rat) 315346, (Mouse) 16402, (Rat) 24511, (Mouse) 16412
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support