Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 12
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: ANRCKKAPV KSCTECVRVD KDCAYCTDEM FRDRRCNTQA ELLAAGCQRE SIVVMESSFQ ITEETQIDTT LRRSQMSPQG LRVRLRPGEE RHFELEVFEP LESPVDLYIL MDFSNSMSDD LDNLKKMGQN LARVLSQLTS DYTIGFGKFV DKVSVPQTDM RPEKLKEPWP NSDPPFSFKN VISLTEDVDE FRNKLQGERI SGNLDAPEGG FDAILQTAVC TRDIGWRPDS THLLVFSTES AFHYEADGAN VLAGIMSRND ERCHLDTTGT YTQYRTQDYP SVPTLVRLLA KHNIIPIFAV TNYSYSYYEK LHTYFPVSSL GVLQEDSSN (27-354 aa encoded by BC126411)
Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha 6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD antigen CD104; CD104 antigen; CD104a; GP150; unnamed protein product
Gene Aliases: C230078O20; CD104; GP150; JEB5A; JEB5B
UniProt ID: (Human) P16144, (Mouse) A2A863, (Rat) Q64632
Entrez Gene ID: (Human) 3691, (Mouse) 192897, (Rat) 25724
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support