Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: Cytoplasm.Cell membrane. Mitochondrion. Nucleus. Secreted.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Lysyl-tRNA synthetase is a homodimer localized to the cytoplasm which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 6.1.1.6; lysine tRNA ligase; Lysine--tRNA ligase; LysRS; Lysyl-tRNA synthetase
Gene Aliases: CMTRIB; DFNB89; KARS; KARS1; KARS2; KIAA0070; KRS
UniProt ID: (Human) Q15046
Entrez Gene ID: (Human) 3735
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support