Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KCNQ1 Antibodies

          Alomone Labs, LTD

          KCNQ1 Polyclonal Antibody

          View all (25) KCNQ1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite KCNQ1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          KCNQ1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          KCNQ1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KCNQ1 Antibody (APC-022-GP200UL) in IHC (F)

          Expression of KCNQ1 in mouse heart - Immunohistochemical staining of mouse heart frozen section using Guinea pig Anti-KCNQ1 Antibody (#APC-022-GP), (1:200). A. KCNQ1 staining (red) appears in muscle fiber membranes (arrows). B. Nuclei are stained using DAPI counterstain (blue). C. Merged image of panels A and B. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KCNQ1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          KCNQ1 Antibody in Western Blot (WB)
          KCNQ1 Polyclonal Antibody

          Product Details

          APC-022-GP200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Guinea pig / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          (C)TYEQLTVPRRGPDEGS, corresponding to amino acid residues 661-676 of human KCNQ1, Intracellular, C-terminus
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.8 mg/mL

          Amount

          160 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. A specific K+ channel, comprised of an alpha subunit KCNQ1 and a beta subunit KCNE1, a small protein which spans the membrane only once, is predominantly expressed in the heart and in the cochlea, and is responsible for regulating the slow, depolarization-activated potassium current. Mutations in the genes encoding for KCNQ1 and KCNE1 lead to cardiac disease because they directly impair electrical signaling, and mutations in KCNQ4 are implicated in the onset of deafness. KCNQ proteins, including KCNQ1 and KCNQ4, characteristically contain six transmembrane domains and function as tetramers. KCNQ4 forms heteromeric channels with KCNQ3 and is expressed in several tissues, including the cochlea, where it is present in outer hair cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ26167; IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1; kidney and cardiac voltage dependend K+ channel; KQT-like 1; potassium channel, voltage gated KQT-like subfamily Q, member 1; Potassium voltage-gated channel subfamily KQT member 1; potassium voltage-gated channel, KQT-like subfamily, member 1; potassium voltage-gated channel, subfamily Q, member 1; slow delayed rectifier channel subunit; tKvLQT1; unnamed protein product; Voltage-gated potassium channel subunit Kv7.1

          View more View less

          Gene Aliases: ATFB1; ATFB3; AW559127; JLNS1; KCNA8; KCNA9; KCNQ1; Kv1.9; Kv7.1; KVLQT1; LQT; LQT1; RWS; SQT2; WRS

          View more View less

          UniProt ID: (Human) P51787, (Mouse) P97414, (Rat) Q9Z0N7

          View more View less

          Entrez Gene ID: (Human) 3784, (Mouse) 16535, (Rat) 84020

          View more View less

          Function(s)
          ion channel activity voltage-gated potassium channel activity delayed rectifier potassium channel activity potassium channel activity protein binding calmodulin binding phosphatidylinositol-4,5-bisphosphate binding protein phosphatase 1 binding outward rectifier potassium channel activity ubiquitin protein ligase binding protein kinase A catalytic subunit binding protein kinase A regulatory subunit binding ion channel binding voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization voltage-gated potassium channel activity involved in atrial cardiac muscle cell action potential repolarization scaffold protein binding voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization voltage-gated ion channel activity protein homodimerization activity voltage-gated ion channel
          Process(es)
          action potential gastric acid secretion gastrin-induced gastric acid secretion regulation of heart rate system process glucose metabolic process ion transport potassium ion transport sodium ion transport heart development sensory perception of sound rhythmic behavior regulation of heart contraction regulation of blood pressure positive regulation of heart rate iodide transport digestive system process erythrocyte differentiation cellular chloride ion homeostasis response to insulin ion transmembrane transport social behavior corticosterone secretion regulation of membrane potential inner ear morphogenesis inner ear development intestinal absorption detection of mechanical stimulus involved in sensory perception of sound regulation of transport chloride ion homeostasis potassium ion homeostasis transmembrane transport cardiac muscle contraction auditory receptor cell development regulation of membrane repolarization regulation of ventricular cardiac muscle cell membrane repolarization regulation of atrial cardiac muscle cell membrane repolarization positive regulation of cardiac muscle contraction regulation of gastric acid secretion cardiac conduction stomach development renal absorption renal sodium ion absorption cellular response to cAMP cellular response to xenobiotic stimulus potassium ion transmembrane transport cellular response to epinephrine stimulus adrenergic receptor signaling pathway cardiac muscle cell contraction ventricular cardiac muscle cell action potential membrane repolarization membrane repolarization during action potential membrane repolarization during cardiac muscle cell action potential atrial cardiac muscle cell action potential regulation of heart rate by cardiac conduction cochlea development potassium ion export across plasma membrane membrane repolarization during atrial cardiac muscle cell action potential membrane repolarization during ventricular cardiac muscle cell action potential positive regulation of potassium ion transmembrane transport negative regulation of delayed rectifier potassium channel activity negative regulation of voltage-gated potassium channel activity non-motile cilium assembly potassium ion import across plasma membrane positive regulation of defense response to virus by host regulation of gene expression by genetic imprinting transport gene silencing regulation of ion transmembrane transport cellular response to drug negative regulation of insulin secretion positive regulation of gastric acid secretion potassium ion export response to anesthetic cardiovascular system development male gonad development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline