Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is specific to LACTB.
Immunogen sequence: DPEASPLAE PPQEQSLAPW SPQTPAPPCS RCFARAIESS RDLLHRIKDE VGAPGIVVGV SVDGKEVWSE GLGYADVENR VPCKPETVMR IASISKSLTM VALAKLWEAG KLDLDIPVQH YVPEFPEKEY EGEKVSVTTR LLISHLSGIR HYEKDIKKVK EEKAYKALKM MKENVAFEQE KEGKSNEKND FTKFKTEQEN EAKCRNSKPG KKKNDFEQGE LYLREKFENS IESLRLFKND PLFFKPGSQF LYSTFGYTLL AAIVERASGC KYLDYMQKIF HDLDMLTTVQ EENEPVIYNR AR (73-373 aa encoded by BC067288)
This gene encodes a protein from the large 39S subunit of the mitochondrial ribosome (mitoribosome). The encoded protein has some sequence similarity to prokaryotic beta-lactamases but most of the residues that are responsible for the beta-lactamase activity are not conserved between the two proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochondrial 39S ribosomal protein L56; serine beta-lactamase-like protein, mitochondrial; unnamed protein product
Gene Aliases: G24; MRPL56
UniProt ID: (Human) P83111
Entrez Gene ID: (Human) 114294
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support