Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • LONP1 Antibodies

          Abnova

          LONP1 Polyclonal Antibody, MaxPab™

          View all (8) LONP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LONP1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          LONP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          LONP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LONP1 Antibody (H00009361-D01P) in ICC/IF

          Immunofluorescence of purified MaxPab antibody to LONP1 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LONP1 Antibody in Immunocytochemistry (ICC/IF)
          LONP1 Antibody in Western Blot (WB)
          LONP1 Antibody in Western Blot (WB)
          LONP1 Antibody in Western Blot (WB)
          LONP1 Antibody in Western Blot (WB)
          LONP1 Polyclonal Antibody, MaxPab™

          Product Details

          H00009361-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          LONP1 (NP_004784.2, 1 a.a. approximately 959 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MAASTGYVRL WGAARCWVLR RPMLAAAGGR VPTAAGAWLL RGQRTCDASP PWALWGRGPA IGGQWRGFWE ASSRGGGAFS GGEDASEGGA EEGAGGAGGS AGAGEGPVIT ALTPMTIPDV FPHLPLIAIT RNPVFPRFIK IIEVKNKKLV ELLRRKVRLA QPYVGVFLKR DDSNESDVVE SLDEIYHTGT FAQIHEMQDL GDKLRMIVMG HRRVHISRQL EVEPEEPEAE NKHKPRRKSK RGKKEAEDEL SARHPAELAM EPTPELPAEV LMVEVENVVH EDFQVTEEVK ALTAEIVKTI RDIIALNPLY RESVLQMMQA GQRVVDNPIY LSDMGAALTG AESHELQDVL EETNIPKRLY KALSLLKKEF ELSKLQQRLG REVEEKIKQT HRKYLLQEQL KIIKKELGLE KDDKDAIEEK FRERLKELVV PKHVMDVVDE ELSKLGLLDN HSSEFNVTRN YLDWLTSIPW GKYSNENLDL ARAQAVLEED HYGMEDVKKR ILEFIAVSQL RGSTQGKILC FYGPPGVGKT SIARSIARAL NREYFRFSVG GMTDVAEIKG HRRTYVGAMP GKIIQCLKKT KTENPLILID EVDKIGRGYQ GDPSSALLEL LDPEQNANFL DHYLDVPVDL SKVLFICTAN VTDTIPEPLR DRMEMINVSG YVAQEKLAIA ERYLVPQARA LCGLDESKAK LSSDVLTLLI KQYCRESGVR NLQKQVEKVL RKSAYKIVSG EAESVEVTPE NLQDFVGKPV FTVERMYDVT PPGVVMGLAW TAMGGSTLFV ETSLRRPQDK DAKGDKDGSL EVTGQLGEVM KESARIAYTF ARAFLMQHAP ANDYLVTSHI HLHVPEGATP KDGPSAGCTI VTALLSLAMG RPVRQNLAMT GEVSLTGKIL PVGGIKEKTI AAKRAGVTCI VLPAENKKDF YDLAAFITEG LEVHFVEHYR EIFDIAFPDE QAEALAVER

          Target Information

          This gene encodes a mitochondrial matrix protein that belongs to the Lon family of ATP-dependent proteases. This protein mediates the selective degradation of misfolded, unassembled or oxidatively damaged polypeptides in the mitochondrial matrix. It may also have a chaperone function in the assembly of inner membrane protein complexes, and participate in the regulation of mitochondrial gene expression and maintenance of the integrity of the mitochondrial genome. Decreased expression of this gene has been noted in a patient with hereditary spastic paraplegia. Alternatively spliced transcript variants have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: h LonHS; hLON ATP-dependent protease; Lon protease homolog, mitochondrial; Lon protease-like protein; LONHs; LONP; Mitochondrial ATP-dependent protease Lon; mitochondrial lon protease-like protein; protease, serine, 15; Serine protease 15; unnamed protein product

          View more View less

          Gene Aliases: 1200017E13Rik; CODASS; hLON; LON; LonHS; LONP; LONP1; PIM1; PRSS15

          View more View less

          UniProt ID: (Human) P36776, (Mouse) Q8CGK3

          View more View less

          Entrez Gene ID: (Human) 9361, (Mouse) 74142

          View more View less

          Function(s)
          nucleotide binding mitochondrial RNA polymerase regulatory region DNA binding DNA binding single-stranded DNA binding single-stranded RNA binding ATP-dependent peptidase activity serine-type endopeptidase activity protein binding ATP binding peptidase activity serine-type peptidase activity hydrolase activity ATPase activity PH domain binding identical protein binding ADP binding insulin receptor substrate binding sequence-specific DNA binding G-quadruplex DNA binding DNA polymerase binding mitochondrial light strand promoter anti-sense binding mitochondrial heavy strand promoter anti-sense binding mitochondrial light strand promoter sense binding mitochondrial heavy strand promoter sense binding serine protease
          Process(es)
          mitochondrial genome maintenance response to hypoxia proteolysis misfolded or incompletely synthesized protein catabolic process mitochondrion organization response to hormone response to aluminum ion protein catabolic process mitochondrial DNA metabolic process cellular response to oxidative stress mitochondrial protein catabolic process negative regulation of insulin receptor signaling pathway chaperone-mediated protein complex assembly proteolysis involved in cellular protein catabolic process protein-containing complex assembly oxidation-dependent protein catabolic process cellular protein complex assembly protein homooligomerization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0