Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey (94%) Mouse, Rat, Hamster, Rabbit (88%) Bat (81%).
The protein encoded by this gene belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. This gene aligns with an internal intron of the retinoblastoma susceptibility gene in the reverse orientation. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled purinergic receptor P2Y5; G-protein coupled receptor; LPA receptor 6; LPA6 Receptor; oleoyl-L-alpha-lysophosphatidic acid receptor; P2Y purinoceptor 5; purinergic receptor 5; purinergic receptor P2Y G protein-coupled protein 5; purinergic receptor P2Y, G-protein coupled, 5; RB intron encoded G-protein coupled receptor; unnamed protein product
Gene Aliases: ARWH1; HYPT8; LAH3; LPA-6; P2RY5; P2Y5
UniProt ID: (Human) P43657
Entrez Gene ID: (Human) 10161
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support