Product References
Myeloid cell expressed proprotein convertase FURIN attenuates inflammation.
Oncotarget
Cordova ZM,Grönholm A,Kytölä V,Taverniti V,Hämäläinen S,Aittomäki S,Niininen W,Junttila I,Ylipää A,Nykter M,Pesu M
RM3004 was used in flow cytometry to investigate the role of FURIN in LysM positive myeloid cells in vivo
Tue Aug 23 00:00:00 EDT 2016
Stromal senescence establishes an immunosuppressive microenvironment that drives tumorigenesis.
Nature communications
Ruhland MK,Loza AJ,Capietto AH,Luo X,Knolhoff BL,Flanagan KC,Belt BA,Alspach E,Leahy K,Luo J,Schaffer A,Edwards JR,Longmore G,Faccio R,DeNardo DG,Stewart SA
RM3004 was used in immunohistochemistry to test if senescent stromal cells contribute to tumorigenesis
Wed Jun 08 00:00:00 EDT 2016
PLD-Specific Small-Molecule Inhibitors Decrease Tumor-Associated Macrophages and Neutrophils Infiltration in Breast Tumors and Lung and Liver Metastases.
PloS one
Henkels KM,Muppani NR,Gomez-Cambronero J
RM3004 was used in immunohistochemistry - paraffin section to assess the importance of targeting phospholipase D-2 on the breast cancer tumor microenvironment
Mon Jun 26 00:00:00 EDT 2017
Interleukin-10 From Marginal Zone Precursor B-Cell Subset Is Required for Costimulatory Blockade-Induced Transplantation Tolerance.
Transplantation
Lal G,Nakayama Y,Sethi A,Singh AK,Burrell BE,Kulkarni N,Brinkman CC,Iwami D,Zhang T,Bromberg JS
RM3004 was used in flow cytometry and immunohistochemistry to determine if specific B cell subsets or if B cell-derived interleukin-10 contributes to tolerance
Tue Sep 01 00:00:00 EDT 2015
Absence of IL-23p19 in donor allogeneic cells reduces mortality from acute GVHD.
Bone marrow transplantation
Thompson JS,Chu Y,Glass JF,Brown SA
RM3004 was used in flow cytometry to test if deficiency of p19 in the allogeneic donor transplant reduces the inflammation caused by acute GVHD
Thu Apr 01 00:00:00 EDT 2010
The manganese superoxide dismutase mimetic, M40403, protects adult mice from lethal total body irradiation.
Free radical research
Thompson JS,Chu Y,Glass J,Tapp AA,Brown SA
RM3004 was used in flow cytometry to report that a non-peptidyl mimetic of MnSOD reduces total body irradiation-induced tissue damage
Sat May 01 00:00:00 EDT 2010
Thymic alterations in mice deficient for the SNARE protein VAMP8/endobrevin.
Cell and tissue research
Kanwar N,Fayyazi A,Backofen B,Nitsche M,Dressel R,von Mollard GF
RM3004 was used in flow cytometry to elucidate the role of VAMP8 in the thymus
Sat Nov 01 00:00:00 EDT 2008
Characterization of early gamma interferon (IFN-gamma) expression during murine listeriosis: identification of NK1.1+ CD11c+ cells as the primary IFN-gamma-expressing cells.
Infection and immunity
Chang SR,Wang KJ,Lu YF,Yang LJ,Chen WJ,Lin YH,Chang HH,Wang SL
RM3004 was used in flow cytometry to identify the cells that are producing gamma interferon during infection with Listeria monocytogenes
Thu Mar 01 00:00:00 EST 2007
Hematopoiesis and stem cell renewal in long-term bone marrow cultures containing catalase.
Blood
Gupta R,Karpatkin S,Basch RS
RM3004 was used in flow cytometry to investigate how peroxide affects hematopoiesis
Wed Mar 01 00:00:00 EST 2006
Dendritic cell differentiation potential of mouse monocytes: monocytes represent immediate precursors of CD8- and CD8+ splenic dendritic cells.
Blood
León B,Martínez del Hoyo G,Parrillas V,Vargas HH,Sánchez-Mateos P,Longo N,López-Bravo M,Ardavín C
RM3004 was used in flow cytometry to discuss DC generation from monocytes.
Thu Apr 01 00:00:00 EST 2004
Regulation of thymus size by competition for stromal niches among early T cell progenitors.
Journal of immunology (Baltimore, Md. : 1950)
Prockop SE,Petrie HT
RM3004 was used in flow cytometry to investigate mechanisms that regulate thymus size
Sun Aug 01 00:00:00 EDT 2004