    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • O-GlcNAc Antibodies
          Invitrogen
          O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal Antibody (9D1.E4)
          1 Published Figure
          1 Reference
          View all (11) O-GlcNAc antibodies
          Questions & Answers
          Cite O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal Antibody (9D1.E4)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          • Published Figures (1)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody (MA1-039) in WB
          Western blot analysis of O-GlcNAc was performed by loading 10 µg of HEK293T or C2C12 whole cell lysates onto a 4-20% Tris-HCl polyacrylamide gel. Proteins were transferred to a PVDF membrane and blocked with 5% BSA/TBST for at least 1 hour. Membranes were then probed with a mouse monoclonal antibody recognizing O-GlcNAc (Product # MA1-039) at a dilution of 1:1000 overnight at 4°C on a rocking platform. Memb... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Immunoprecipitation (IP)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          Product Details
          MA1-039
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          View 1 publication 1 publication
          Immunoprecipitation (IP)
          5 µg
          -
          Product Specifications
          Species Reactivity
          Chemical
          Published species
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          9D1.E4
          Immunogen
          SANM-conjugated linear peptide corresponding to human O-GluNAc.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS with 1mg/mL BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536726
          Product Specific Information
          MA1-039 has been successfully used in Western blot and immunoprecipitation applications with human samples. In Western blot, MA1-039 detects proteins with O-GlcNAc post-translational modification, and therefore will detect multiple specific bands.
          Target Information
          O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.
          Bioinformatics
          Protein Aliases: GlcNAc
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline