    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Adenovirus Hexon Antibodies
          Invitrogen
          Adenovirus Hexon Monoclonal Antibody (D36J), FITC
          View all (11) Adenovirus Hexon antibodies
          Questions & Answers
          Cite Adenovirus Hexon Monoclonal Antibody (D36J), FITC
          Additional Information:
          {{banner.description}}
          Product Details
          MA1-7329
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:5-1:20
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Virus
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          D36J
          Immunogen
          Adenovirus hexon protein.
          Conjugate
          FITC FITC FITC
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          498/517 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2, with 10mg/mL BSA
          Contains
          0.1% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1016582
          Product Specific Information
          The MA1-7329 antibody reacts with Adenovirus hexon in viral and infected tissue samples.
          MA1-7329 has been successfully used in ELISA and Immunofluorescence applications.
          The MA1-7329 antibody was raised against Adenovirus hexon protein.
          Target Information
          Hexon protein is a major coat protein of adenoviruses. Adenoviruses capsids have three principal protein components: the hexon, the penton, and the fiber. Hexon consists of three subunits together forming two major components of different morphological symmetry. A triangular top with three towers of density is superimposed on a more bulky pseudo hexagonal base.
          Bioinformatics
          Protein Aliases: Adeno Virus; AdV; HADV; HAdVC_gp08
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline