    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Mycobacterium tuberculosis HSP65 Antibodies
          Invitrogen
          Mycobacterium tuberculosis HSP65 Monoclonal Antibody (J105G)
          View all (2) Mycobacterium tuberculosis HSP65 antibodies
          Questions & Answers
          Cite Mycobacterium tuberculosis HSP65 Monoclonal Antibody (J105G)
          Additional Information:
          {{banner.description}}
          Product Details
          MA1-7421
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:10-1:50
          -
          ELISA (ELISA)
          1:20-1:200
          -
          Product Specifications
          Species Reactivity
          Bacteria
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          J105G
          Immunogen
          M. tuberculosis Heat shock protein 65.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2
          Contains
          0.1% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1017676
          Product Specific Information
          The MA1-7421 antibody reacts with Mycobacter. tuberculosis HSP65. Non-reactive with other Mycobacteria.
          MA1-7421 has been successfully used in ELISA, and Western blot applications.
          The MA1-7421 antibody was raised against M. tuberculosis Heat shock protein 65.
          Target Information
          Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
          Bioinformatics
          Protein Aliases: m tuberculosis; M. tuberculosis
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline