    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • V5 Tag Antibodies
          Invitrogen
          V5 Tag Monoclonal Antibody (SV5-Pk1), FITC
          View all (83) V5 Tag antibodies
          Questions & Answers
          Cite V5 Tag Monoclonal Antibody (SV5-Pk1), FITC
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          V5 Tag Antibody (MA1-80281) in ICC/IF
          Immunofluorescent analysis of V5 Tag was performed using 70% confluent V5-H3-6XHis transfected HEK-293 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with V5 Tag Monoclonal Antibody (SV5-Pk1), FITC (Product # MA1-80281) at 1:100 dilution and Histone H3 Recombinant Polyclonal Antibody (Product # 711055) at 0.5 µg/mL in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled wit... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          MA1-80281
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          SV5-Pk1
          Immunogen
          Paramyxovirus SV5, Simian-Virus 5.
          Conjugate
          FITC FITC FITC
          Excitation/Emission Max
          498/517 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS with 1% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_935873
          Product Specific Information
          MA1-80281 recognizes the sequence IPNPLLGL, present on the P/V proteins of the paramyxovirus, SV5. This antibody has been used to detect recombinant proteins, including epitope-tagged transmembrane and secreted proteins. Successfully used tags include a 14-amino acid tag (gly lys pro ile pro asn pro leu leu gly leu asp ser thr) and a smaller 9-amino acid tag that has been used to inhibit the binding of the monoclonal antibody to its native protein.
          Target Information
          Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).
          Bioinformatics
          Protein Aliases: simian virus 5 antibody; SPV5gp2; sv5; v-5; V5; V5 Epitope Tag
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline