    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • S-peptide Epitope Tag Antibodies
          Invitrogen
          S-peptide Epitope Tag Monoclonal Antibody (6.2)
          7 Published Figures
          12 References
          View all (15) S-peptide Epitope Tag antibodies
          Questions & Answers
          Cite S-peptide Epitope Tag Monoclonal Antibody (6.2)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (7)
          S-peptide Epitope Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          S-peptide Epitope Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 8
          S-peptide Epitope Tag Antibody (MA1-981) in ICC/IF
          Immunofluorescence of S-peptide tagged topisomerase I in the nuclei of transfected K562 cells using Product # MA1-981. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          S-peptide Epitope Tag Antibody in Immunocytochemistry (ICC/IF)
          S-peptide Epitope Tag Antibody in Western Blot (WB)
          S-peptide Epitope Tag Antibody in Western Blot (WB)
          S-peptide Epitope Tag Antibody in Western Blot (WB)
          S-peptide Epitope Tag Antibody in Western Blot (WB)
          S-peptide Epitope Tag Antibody in Western Blot (WB)
          S-peptide Epitope Tag Antibody in Western Blot (WB)
          S-peptide Epitope Tag Antibody in Western Blot (WB)
          Product Details
          MA1-981
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1 µg/mL
          View 10 publications 10 publications
          Immunohistochemistry (IHC)
          10 µg/mL
          View 1 publication 1 publication
          Immunocytochemistry (ICC/IF)
          10 µg/mL
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Tag
          Published species
          Tag
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          6.2
          Immunogen
          Synthetic Peptide: K(1) E T A A A K F E R Q H M D S(15) C
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS with 1mg/mL BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_347008
          Product Specific Information
          This antibody detects the S-peptide in human samples.
          MA1-981 has been successfully used in Western blot, immunohistochemistry and immunofluorescence procedures. By Western blot this antibody detects the S-peptide in K562 cells. In immunohistochemistry, and immunofluorescence procedures MA1-981 recognizes the S-peptide in transfected K562 cells.
          The MA1-981 immunogen corresponds to the S-peptide conjugated through a C-terminal cysteine to keyhole limpet hemocyanin.
          Target Information
          The S-Peptide epitope tag (S-Tag (TM)) can be used for numerous applications which include the analysis of localization, expression detection, and purification of polypeptides from mammalian cells. The small size of the tag (15 aa), the ability to purify tagged polypeptides without antibodies, and the very low expression of endogenous tissues in mammalian cells that react with S-Peptide make the S-Peptide antibody an attractive alternative to many existing epitope tags. Epitope tagging is a powerful and versatile strategy for detecting and purifying proteins expressed by cloned genes. To utilize this feature, protein expression vectors are typically engineered with a nucleotide sequence that encodes the peptide epitope tag. The gene of interest is cloned in-frame relative to the tag and, upon expression, the protein of interest is synthesized as a fusion protein with the peptide tag. Fusion protein detection and/or purification is mediated by highly specific antibodies to the engineered peptide, thus eliminating the need for antibodies to proteins from each newly cloned gene.
          Bioinformatics
          Protein Aliases: S-tag
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline