    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • beta Actin Antibodies
          Invitrogen
          beta Actin Loading Control Monoclonal Antibody (BA3R)
          Advanced Verification
          82 Published Figures
          477 References
          View all (99) beta Actin antibodies
          Questions & Answers
          Cite beta Actin Loading Control Monoclonal Antibody (BA3R)
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (12)
          • Published Figures (82)
          • Advanced Verification (1)
          beta Actin Loading Control Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          beta Actin Loading Control Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 95
          beta Actin Loading Control Antibody (MA5-15739) in ICC/IF
          Immunofluorescence analysis of beta Actin was performed using 70% confluent log phase LNCaP cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with beta Actin (BA3R) Loading Control Mouse Monoclonal Antibody (Product # MA5-15739-1MG) at 2µg/mL in 0.1% BSA and... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          beta Actin Loading Control Antibody in Immunocytochemistry (ICC/IF)
          beta Actin Loading Control Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Actin Loading Control Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Actin Loading Control Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Flow Cytometry (Flow)
          beta Actin Loading Control Antibody in Flow Cytometry (Flow)
          beta Actin Loading Control Antibody in Flow Cytometry (Flow)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          beta Actin Loading Control Monoclonal Antibody (BA3R)
          Product Details
          MA5-15739
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:10,000
          View 441 publications 441 publications
          Immunohistochemistry (IHC)
          -
          View 5 publications 5 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:100-1:1,000
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          View 19 publications 19 publications
          Flow Cytometry (Flow)
          1-5 µg/1x10^6 cells
          View 1 publication 1 publication
          ELISA (ELISA)
          Assay-dependent
          View 1 publication 1 publication
          Immunoprecipitation (IP)
          -
          View 2 publications 2 publications
          Control (Ctrl)
          -
          View 1 publication 1 publication
          Inhibition Assays (Inhibition)
          -
          View 1 publication 1 publication
          Miscellaneous PubMed (Misc)
          -
          View 6 publications 6 publications
          Product Specifications
          Species Reactivity
          Chicken, Human, Mouse, Rabbit, Rat
          Published species
          Bacteria, Chicken, Cynomolgus monkey, Dog, Fungi, Guinea pig, Hamster, Human, Mouse, Non-human primate, Pig, Plant, Rabbit, Rat, Xenopus
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          BA3R
          Immunogen
          Beta-actin N-terminal peptide.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • DyLight 488
          • DyLight 550
          • DyLight 650
          • DyLight 680
          • DyLight 800 4X PEG
          • HRP
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2
          Contains
          no preservative
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_10979409
          Product Specific Information
          Storage and handling: add 0.05% sodium azide if desired. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Target Information
          Beta actin is one of six actin isoforms which have been identified. Beta actin is a non-muscle cytoskeletal protein in all human cell types and is involved in cell motility, structure, and integrity. There are six different actin isoforms in human. The beta isoform of actin, along with gamma actin, coexist in most cell types as components of the cyto-skeleton. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Because beta actin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting. Antibodies to beta-actin provide a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.
          Bioinformatics
          Protein Aliases: A-X actin; A-X actin-like protein; Actin is a protein with a high degree of evolutionary conservation that undergoes polymerization to generate filaments, which subsequently assemble into interconnected networks within the cytoplasm of cells.; actin, beta, cytoplasmic; Bact; actin; beta cytoskeletal actin; beta-actin; cytoplasmic 1; cytoplasmic beta-actin; cytoplasmic protein; gamma non-muscle actin; I(2)-actin; melanoma X-actin; PS1TP5-binding protein 1; unnamed protein product
          View more View less
          Gene Aliases: ACT-5; actin; Actx; Bact; beta-actin; BKRNS; BNS; BRWS1; CSMH; DDS1; E430023M04Rik; PS1TP5BP1; THC8
          View more View less
          UniProt ID: (Rabbit) P29751, (Human) P60709, (Chicken) P60706, (Mouse) P60710, (Rat) P60711
          View more View less
          Entrez Gene ID: (Rabbit) 100009272, (Human) 60, (Chicken) 396526, (Mouse) 11461, (Rat) 81822
          View more View less
          Function(s)
          structural constituent of cytoskeleton protein binding ATPase activity kinesin binding protein kinase binding nitric-oxide synthase regulator activity Tat protein binding nucleosomal DNA binding identical protein binding tau protein binding nitric-oxide synthase binding structural constituent of postsynaptic actin cytoskeleton transporter regulator activity ATP binding actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          morphogenesis of a polarized epithelium chromatin remodeling regulation of transcription from RNA polymerase II promoter cytoskeleton organization microtubule nucleation establishment or maintenance of cell polarity axonogenesis positive regulation of cell proliferation substantia nigra development regulation of mitotic metaphase/anaphase transition adherens junction assembly maintenance of blood-brain barrier regulation of apoptotic process apical protein localization positive regulation of T cell differentiation negative regulation of cell differentiation positive regulation of cell differentiation positive regulation of myoblast differentiation positive regulation of transcription, DNA-templated cell motility regulation of norepinephrine uptake positive regulation of norepinephrine uptake regulation of cell cycle regulation of G0 to G1 transition platelet aggregation protein localization to adherens junction cellular response to cytochalasin B postsynaptic actin cytoskeleton organization regulation of transepithelial transport regulation of synaptic vesicle endocytosis positive regulation of stem cell population maintenance positive regulation of double-strand break repair via homologous recombination regulation of G1/S transition of mitotic cell cycle regulation of double-strand break repair positive regulation of double-strand break repair regulation of nucleotide-excision repair cellular response to electrical stimulus regulation of protein localization to plasma membrane circadian rhythm response to mechanical stimulus dynein-driven meiotic oscillatory nuclear movement response to immobilization stress retina development in camera-type eye
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b6cf8ccb7-rktn8:80/100.66.72.150:80.
          git-commit: 76b2fce788d0185f5e22c122d85c407206860d42
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.0-2026-offline-prod