    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CHRDL2 Antibodies
          Invitrogen
          CHRDL2 Monoclonal Antibody (328905)
          View all (11) CHRDL2 antibodies
          Questions & Answers
          Cite CHRDL2 Monoclonal Antibody (328905)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CHRDL2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          CHRDL2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CHRDL2 Antibody (MA5-24056) in IHC (F)
          Immunohistochemical analysis of CHRDL2 in immersion fixed frozen sections of embryonic mouse cartilage (15 d.p.c.). Samples were incubated in CHRDL2 monoclonal antibody (Product # MA5-24056) using a dilution of 25 µg/mL overnight at 4 °C. Tissue was stained with the Anti-Rat HRP-DAB Cell & Tissue Staining Kit (brown) and counterstained with hematoxylin (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CHRDL2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          CHRDL2 Monoclonal Antibody (328905)
          Product Details
          MA5-24056
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1 µg/mL
          -
          Immunohistochemistry (Frozen) (IHC (F))
          8-25 µg/mL
          -
          Immunoprecipitation (IP)
          25 µg/mL
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Rat / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          328905
          Immunogen
          Mouse myeloma cell line NS0-derived recombinant mouse Chordin-like 2/CHRDL2 Gln24-Leu426
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS with 5% trehalose
          Contains
          No Preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2606392
          Product Specific Information
          In direct ELISAs and Western blots, no cross-reactivity with recombinant human CHRDL2, recombinant mouse (rm) CHRDL1, or rmChordin is observed.
          Reconstitute at 0.5 mg/mL in sterile PBS.
          Target Information
          CHRDL2 encodes a member of the chordin family of proteins. Chordin family members are secreted proteins that share a cysteine-rich pro-collagen repeat domain and associate with members of the transforming growth factor beta superfamily. In vitro assays demonstrate a direct interaction between the encoded protein and human activin A. This gene is expressed in many tissues including osteoblasts, where it is differentially expressed during differentiation. In addition, its expression is upregulated in human osteoarthritic joint cartilage, suggesting a role in adult cartilage regeneration.
          Bioinformatics
          Protein Aliases: chordin like 2; Chordin-like protein 2; FKSG37
          View more View less
          Gene Aliases: 1810022C01Rik; Chl2; Chrdl2
          View more View less
          UniProt ID: (Mouse) Q8VEA6
          View more View less
          Entrez Gene ID: (Mouse) 69121
          View more View less
          Function(s)
          protein binding
          Process(es)
          ossification multicellular organism development cell differentiation negative regulation of BMP signaling pathway cartilage development
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-b6zwj:80/100.66.74.164:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline