    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cytokeratin 14 Antibodies
          Invitrogen
          Cytokeratin 14 Monoclonal Antibody (LL002), FITC
          View all (114) Cytokeratin 14 antibodies
          Questions & Answers
          Cite Cytokeratin 14 Monoclonal Antibody (LL002), FITC
          Additional Information:
          {{banner.description}}
          Cytokeratin 14 Monoclonal Antibody (LL002), FITC
          Product Details
          MA5-28125
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          Flow Cytometry (Flow)
          Neat-1:10
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG3
          Class
          Monoclonal
          Type
          Antibody
          Clone
          LL002
          Immunogen
          Last 15 C-terminal residues of human cytokeratin 14 conjugated to thyroglobulin
          3D Epitope / Immunogen
          Conjugate
          FITC FITC FITC
          Excitation/Emission Max
          498/517 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS with 1% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2745108
          Target Information
          Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (ie. skin) and non-keratinizing cells (ie. mesothelial cells). At least 20 different CKs can be identified. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). Belonging to the type I subfamily of low molecular weight keratins and existing in combination with keratin 5, keratin 14 distinguishes stratified epithelial cells from simple epithelial cells and is useful in identification of squamous cell carcinomas. It is considered a prognostic marker in breast carcinomas. At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: CK-14; cytokeratin 14; Cytokeratin-14; K14; keratin 14, type I; Keratin, type I cytoskeletal 14; Keratin-14; Koebner
          View more View less
          Gene Aliases: CK14; EBS1; EBS1A; EBS1B; EBS1C; EBS1D; EBS3; EBS4; K14; KRT14; NFJ
          View more View less
          UniProt ID: (Human) P02533
          View more View less
          Entrez Gene ID: (Human) 3861
          View more View less
          Function(s)
          structural molecule activity structural constituent of cytoskeleton protein binding structural constituent of epidermis keratin filament binding
          Process(es)
          morphogenesis of an epithelium epidermis development response to radiation keratinocyte differentiation hair cycle intermediate filament organization intermediate filament bundle assembly stem cell differentiation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline