    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RIPK4 Antibodies
          Invitrogen
          RIPK4 Monoclonal Antibody (1F7D9)
          View all (14) RIPK4 antibodies
          Questions & Answers
          Cite RIPK4 Monoclonal Antibody (1F7D9)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          RIPK4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RIPK4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          RIPK4 Antibody (MA5-31895) in WB
          Western blot analysis of RIPK4 in HEK293 (1) and RIPK4 cell lysate. Samples were incubated with RIPK4 monoclonal antibody (Product # MA5-31895) using a dilution of 1:500-1:2000. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RIPK4 Antibody in Western Blot (WB)
          RIPK4 Antibody in Western Blot (WB)
          RIPK4 Antibody in Flow Cytometry (Flow)
          RIPK4 Antibody in ELISA (ELISA)
          RIPK4 Monoclonal Antibody (1F7D9)
          Product Details
          MA5-31895
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Flow Cytometry (Flow)
          1:200-1:400
          -
          ELISA (ELISA)
          1:10,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1F7D9
          Immunogen
          Purified recombinant fragment of human RIPK4 (AA: 675-784) expressed in E. coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS
          Contains
          0.05% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2787518
          Product Specific Information
          MA5-31895 has been tested in indirect ELISA.
          Target Information
          The protein encoded by this gene is a serine/threonine protein kinase that interacts with protein kinase C-delta. The encoded protein can also activate NFkappaB and is required for keratinocyte differentiation. This kinase undergoes autophosphorylation.
          Bioinformatics
          Protein Aliases: ankyrin repeat domain-containing protein 3; PKC-delta-interacting protein kinase; protein kinase C-associated kinase; serine/threonine-protein kinase ANKRD3; unnamed protein product
          View more View less
          Gene Aliases: ANKK2; ANKRD3; CHANDS; DIK; NKRD3; PKK; PPS2; RIP4
          View more View less
          Entrez Gene ID: (Human) 54101
          View more View less
          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity transferase activity protein serine kinase activity
          Process(es)
          morphogenesis of an epithelium skin development positive regulation of NF-kappaB transcription factor activity
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0