    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Contactin 5 Antibodies
          Invitrogen
          Contactin 5 Monoclonal Antibody (4C10E3C12)
          View all (11) Contactin 5 antibodies
          Questions & Answers
          Cite Contactin 5 Monoclonal Antibody (4C10E3C12)
          Additional Information:
          {{banner.description}}
          Contactin 5 Monoclonal Antibody (4C10E3C12)
          Product Details
          MA5-42992
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          1:1,000-1:2,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4C10E3C12
          Immunogen
          Recombinant Human CNTN5 protein (1-1059).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          no preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2912133
          Product Specific Information
          Specificity: Human CNTN5/Contactin-5
          Target Information
          The protein encoded by this gene is a member of the immunoglobulin superfamily, and contactin family, which mediate cell surface interactions during nervous system development. This protein is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
          Bioinformatics
          Protein Aliases: Contactin-5; hNB-2; MGC163491; neural adhesion molecule; Neural recognition molecule NB-2; unnamed protein product
          View more View less
          Gene Aliases: CNTN5; HNB-2s; NB-2
          View more View less
          UniProt ID: (Human) O94779
          View more View less
          Entrez Gene ID: (Human) 53942
          View more View less
          Function(s)
          protein binding involved in cell-cell adhesion
          Process(es)
          cell adhesion axon guidance brain development sensory perception of sound cell-cell adhesion presynapse assembly
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline