    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Ceftiofur Antibodies
          Invitrogen
          Ceftiofur Monoclonal Antibody (3E2)
          Questions & Answers
          Cite Ceftiofur Monoclonal Antibody (3E2)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Ceftiofur Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          Ceftiofur Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Ceftiofur Antibody (MA5-49721) in SDS-PAGE
          SDS-PAGE analysis of Ceftiofur monoclonal antibody (Product # MA5-49721). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Ceftiofur Antibody in SDS-PAGE (SDS-PAGE)
          Product Details
          MA5-49721
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (IHC)
          Assay-dependent
          -
          ELISA (ELISA)
          1:5,000
          -
          Lateral Flow (LF)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Chemical
          Host/Isotype
          Mouse / IgG
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3E2
          Immunogen
          Conjugate of CEF hapten and KLH
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.42 mg/mL
          Amount
          200 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.4
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3090215
          Product Specific Information
          Antibody tested in indirect ELISA
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline