    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Digoxigenin Antibodies
          Invitrogen
          Digoxigenin Recombinant Rabbit Monoclonal Antibody (PSH04-39)
          View all (8) Digoxigenin antibodies
          Questions & Answers
          Cite Digoxigenin Recombinant Rabbit Monoclonal Antibody (PSH04-39)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          Digoxigenin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Digoxigenin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Digoxigenin Antibody (MA5-56506) in IHC (P)
          Immunohistochemistry analysis of Digoxigenin with paraffin-embedded mouse spleen tissue. The tissues were blocked in 1% BSA (20 min, room temp), washed with ddH2O and PBS, incubated with Digoxigenin Recombinant Monoclonal Antibody (Product # MA5-56506) at a dilution of 1:1,000 (1 hr, room temp). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Digoxigenin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Digoxigenin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Digoxigenin Antibody in Western Blot (WB)
          Product Details
          MA5-56506
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200-1:5,000
          -
          Immunohistochemistry (Frozen) (IHC (F))
          1:50-1:1,000
          -
          ELISA (ELISA)
          1:5,000-1:20,000
          -
          Immunoprecipitation (IP)
          1-2 µg per sample
          -
          Dot blot (DB)
          1:2,000
          -
          Product Specifications
          Species Reactivity
          Chemical
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          PSH04-39
          Immunogen
          DIG-OVA
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 0.1% BSA, 40% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3679533
          Target Information
          Digoxin, also known as digitalis, is a purified cardiac glycoside extracted from the foxglove plant, Digitalis lanata. Its corresponding aglycone is digoxigenin, and its acetyl derivative is acetyldigoxin. Digoxin is widely used in the treatment of various heart conditions, namely atrial fibrillation, atrial flutter and sometimes heart failure that cannot be controlled by other medication. Digoxin preparations are commonly marketed under the trade names Lanoxin, Digitek, and Lanoxicaps. Digoxin is also used as an affinity system for nonradioactive labeling and detection of proteins and nucleic acids.
          Bioinformatics
          Protein Aliases: C23H34O5; DIG; Digitalis
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline