    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RAP1A/RAP1B Antibodies
          Invitrogen
          RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (24GB1030)
          View all (5) RAP1A/RAP1B antibodies
          Questions & Answers
          Cite RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (24GB1030)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          RAP1A/RAP1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RAP1A/RAP1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          RAP1A/RAP1B Antibody (MA5-57210) in ICC/IF
          Immunocytochemical staining of RAP1A/RAP1B in Hela cells using RAP1A/RAP1B Recombinant Monoclonal Antibody (Product # MA5-57209) at a 1:1,1000 dilution. Nuclei were stained blue with DAPI; RAP1A + RAP1B was stained magenta with Alexa Fluor® 647. Images were taken using Leica stellaris 5. Protein abundance based on laser Intensity and smart gain: Medium. Scale bar: 20 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RAP1A/RAP1B Antibody in Immunocytochemistry (ICC/IF)
          RAP1A/RAP1B Antibody in Western Blot (WB)
          RAP1A/RAP1B Antibody in Flow Cytometry (Flow)
          RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (24GB1030)
          Product Details
          MA5-57210
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:5,000
          -
          Immunocytochemistry (ICC/IF)
          1:100-1:1,000
          -
          Flow Cytometry (Flow)
          1:2,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          24GB1030
          Immunogen
          A synthesized peptide derived from human RAP1A/RAP1B (aa 100-150)
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          RRID
          AB_3680235
          Bioinformatics
          Protein Aliases: C21KG; G-22K; GTP-binding protein smg p21A; GTP-binding protein smg p21B; GTP-binding protein SMG-P21A; NB4 apoptosis/differentiation related protein; RAP1B; Ras family small GTP binding protein RAP1B; RAS related protein 1b; RAS-related protein 1a; Ras-related protein Krev-1; Ras-related protein Rap-1A; Ras-related protein Rap-1b; RAS-related protein RAP1B; small GTP binding protein; unnamed protein product
          View more View less
          Gene Aliases: 2810443E11Rik; AI848598; C21KG; G-22K; K-REV; KREV-1; KREV1; OK/SW-cl.11; RAL1B; rap 1A; RAP-1A; RAP1; RAP1A; RAP1B; SMGP21; THC11
          View more View less
          UniProt ID: (Human) P62834, (Human) P61224, (Mouse) P62835, (Rat) P62836, (Rat) Q62636, (Mouse) Q99JI6
          View more View less
          Entrez Gene ID: (Human) 5906, (Human) 5908, (Mouse) 109905, (Rat) 295347, (Rat) 171337, (Mouse) 215449
          View more View less
          Function(s)
          nucleotide binding GTPase activity obsolete small monomeric GTPase activity guanyl-nucleotide exchange factor activity protein binding GTP binding hydrolase activity GDP binding small GTPase binding macromolecular complex binding protein transporter activity Ras GTPase binding Rap guanyl-nucleotide exchange factor activity protein complex binding small GTPase
          Process(es)
          signal transduction small GTPase mediated signal transduction nervous system development response to carbohydrate positive regulation of neuron projection development synaptic vesicle exocytosis Rap protein signal transduction negative regulation of collagen biosynthetic process nerve growth factor signaling pathway positive regulation of GTPase activity positive regulation of protein kinase activity positive regulation of glucose import positive regulation of phagocytosis positive regulation of Fc receptor mediated stimulatory signaling pathway establishment of endothelial barrier positive regulation of ERK1 and ERK2 cascade cellular response to cAMP protein localization to plasma membrane response to antineoplastic agent liver regeneration regulation of neurotransmitter receptor localization to postsynaptic specialization membrane regulation of cell junction assembly cellular response to forskolin cellular response to nerve growth factor stimulus negative regulation of synaptic vesicle exocytosis positive regulation of vasculogenesis cell proliferation calcium ion regulated exocytosis positive regulation of integrin activation negative regulation of calcium ion-dependent exocytosis establishment of localization in cell modification of postsynaptic structure regulation of establishment of cell polarity protein transport metabolic process microvillus assembly
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline