    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • S6 Antibodies
          Invitrogen
          Phospho-S6 Ribosomal Protein (Ser240, Ser244) Recombinant Rabbit Monoclonal Antibody (7M8E8)
          View all (89) S6 antibodies
          Questions & Answers
          Cite Phospho-S6 Ribosomal Protein (Ser240, Ser244) Recombinant Rabbit Monoclonal Antibody (7M8E8)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Phospho-S6 Ribosomal Protein (Ser240, Ser244) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-S6 Ribosomal Protein (Ser240, Ser244) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Phospho-S6 Ribosomal Protein (Ser240, Ser244) Antibody (MA5-58246) in WB
          Western blot analysis of Phospho-S6 Ribosomal Protein (Ser240, Ser244) in HeLa, NIH/3T3 and PC-12 (30 µg/lane). The cell lines were treated by 10% FBS at 37°C for 30 minutes after serum-starvation overnight. Samples were incubated overnight at 4°C with Phospho-S6 Ribosomal Protein (Ser240, Ser244) Recombinant Monoclonal Antibody (Product # MA5-58246) using a dilution of 1:54, 000. Followed by a HRP-conjugat... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-S6 Ribosomal Protein (Ser240, Ser244) Antibody in Western Blot (WB)
          Phospho-S6 Ribosomal Protein (Ser240, Ser244) Recombinant Rabbit Monoclonal Antibody (7M8E8)
          Product Details
          MA5-58246
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:54,000-1:162,000
          -
          ELISA (ELISA)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          7M8E8
          Immunogen
          A synthetic phosphorylated peptide around S240 & S244 of human RPS6 (NP_001001.2).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.68 mg/mL
          Amount
          68 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.05% BSA, 50% glycerol
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          RRID
          AB_3681266
          Product Specific Information
          Immunogen sequence: STSKS
          Target Information
          Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a cytoplasmic ribosomal protein that is a component of the 40S subunit. The protein belongs to the S6E family of ribosomal proteins. It is the major substrate of protein kinases in the ribosome, with subsets of five C-terminal serine residues phosphorylated by different protein kinases. Phosphorylation is induced by a wide range of stimuli, including growth factors, tumor-promoting agents, and mitogens. Dephosphorylation occurs at growth arrest. The protein may contribute to the control of cell growth and proliferation through the selective translation of particular classes of mRNA. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
          Bioinformatics
          Protein Aliases: 40S ribosomal protein S6; OTTHUMP00000021121; OTTHUMP00000021122; Phosphoprotein NP33; ribosomal protein S6; RP11-513M16.6; RS6; Small ribosomal subunit protein eS6; unnamed protein product
          View more View less
          Gene Aliases: eS6; OK/SW-cl.2; RPS6; S6; S6R
          View more View less
          UniProt ID: (Human) P62753, (Mouse) P62754, (Rat) P62755
          View more View less
          Entrez Gene ID: (Human) 6194, (Mouse) 20104, (Rat) 29304
          View more View less
          Function(s)
          RNA binding mRNA binding structural constituent of ribosome protein binding protein kinase binding poly(A) RNA binding ribosomal protein translational protein
          Process(es)
          ribosomal small subunit assembly cytoplasmic translation rRNA processing translation positive regulation of cell proliferation TOR signaling response to insulin ribosomal small subunit biogenesis glucose homeostasis positive regulation of apoptotic process cellular response to ethanol negative regulation of bicellular tight junction assembly G1/S transition of mitotic cell cycle placenta development T cell proliferation involved in immune response activation-induced cell death of T cells mitotic nuclear division mitotic cell cycle checkpoint gastrulation oogenesis stage T cell differentiation in thymus negative regulation of apoptotic process erythrocyte development
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline