    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DPEP3 Antibodies
          Invitrogen
          DPEP3 Recombinant Mouse Monoclonal Antibody (5H3)
          View all (8) DPEP3 antibodies
          Questions & Answers
          Cite DPEP3 Recombinant Mouse Monoclonal Antibody (5H3)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          DPEP3 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          DPEP3 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          DPEP3 Antibody (MA5-60729) in ELISA
          ELISA analysis of DPEP3. Samples were incubated with DPEP3 Recombinant Monoclonal antibody (Product # MA5-60729) using a dilution of 2 µg/mL. Activity: Measured by its binding ability in a functional ELISA. The EC50 is 7.817-8.936 ng/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DPEP3 Antibody in ELISA (ELISA)
          DPEP3 Antibody in ELISA (ELISA)
          Product Details
          MA5-60729
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          1:5,000-1:50,000
          -
          Product Specifications
          Species Reactivity
          Human, Non-human primate
          Host/Isotype
          Mouse / IgG1
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          5H3
          Immunogen
          Recombinant Human DPEP3 protein.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.03% ProClin 300
          Storage conditions
          -20°C or -80°C if preferred
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3701352
          Target Information
          This gene encodes a membrane-bound glycoprotein from the family of dipeptidases involved in hydrolytic metabolism of various dipeptides, including penem and carbapenem beta-lactam antibiotics. This gene is located on chromosome 16 in a cluster with another member of this family. Alternatively spliced transcript variants that encode different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: Dipeptidase 3; membrane-bound dipeptidase 3; metallopeptidase (family M19); testis secretory sperm-binding protein Li 211a; unnamed protein product
          View more View less
          Gene Aliases: DPEP3; MBD3; UNQ834/PRO1772
          View more View less
          UniProt ID: (Human) Q9H4B8
          View more View less
          Entrez Gene ID: (Human) 64180
          View more View less
          Function(s)
          peptidase activity dipeptidase activity
          Process(es)
          proteolysis leukotriene D4 catabolic process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline