    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Syndecan 2 Antibodies
          Invitrogen
          Syndecan 2 Monoclonal Antibody (305515)
          View all (11) Syndecan 2 antibodies
          Questions & Answers
          Cite Syndecan 2 Monoclonal Antibody (305515)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Syndecan 2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Syndecan 2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Syndecan 2 Antibody (MA5-61235) in Flow
          Flow Cytometry analysis of Syndecan 2 in Human PBMCs using Syndecan 2 monoclonal antibody (Product # MA5-61235). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Syndecan 2 Antibody in Flow Cytometry (Flow)
          Syndecan 2 Monoclonal Antibody (305515)
          Product Details
          MA5-61235
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rat / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          305515
          Immunogen
          Mouse myeloma cell line NS0-derived recombinant human Syndecan-2/CD362, Glu19-Gly144
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS with trehalose
          Contains
          no preservative
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3701847
          Product Specific Information
          Reconstitute at 0.5 mg/mL in sterile PBS.
          Target Information
          Syndecans are heparan sulfate proteoglycans (HSPGs) that are involved in cell adhesion, tissue morphogenesis, differentiation, and act as low affinity co-receptors of soluble growth factors. It is involved in the mitogenic activity and signaling of Granulocyte-Macrophage Colony-stimulating factor in osteoblasts, and has been shown to have a regulatory role in matrix assembly. Syndecan-2 is expressed at epithelial-mesenchymal interfaces, and prechondrogenic and preosteogenic mesenchymal condensations, which are areas of high morphogenetic activity.
          Bioinformatics
          Protein Aliases: CD362; cell surface-associated heparan sulfate proteoglycan 1; Fibroglycan; heparan sulfate proteoglycan 1, cell surface-associated; Heparan sulfate proteoglycan core protein; HSPG; SYND2; syndecan proteoglycan 2; Syndecan-2; unnamed protein product
          View more View less
          Gene Aliases: CD362; HSPG; HSPG1; SDC2; SYND2
          View more View less
          UniProt ID: (Human) P34741
          View more View less
          Entrez Gene ID: (Human) 6383
          View more View less
          Function(s)
          protein binding PDZ domain binding identical protein binding
          Process(es)
          nervous system development cell migration cell differentiation dendrite morphogenesis regulation of dendrite morphogenesis
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline