    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NEDD8 Antibodies
          Invitrogen
          NEDD8 Monoclonal Antibody (866916)
          View all (23) NEDD8 antibodies
          Questions & Answers
          Cite NEDD8 Monoclonal Antibody (866916)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          NEDD8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NEDD8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NEDD8 Antibody (MA5-61500) in WB
          Western Blot analysis of NEDD8 in Raji, NIH-3T3, and NRK using NEDD8 monoclonal antibody (Product # MA5-61500). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NEDD8 Antibody in Western Blot (WB)
          NEDD8 Monoclonal Antibody (866916)
          Product Details
          MA5-61500
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          2 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          8-25 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rat / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          866916
          Immunogen
          E. coli-derived recombinant human NEDD8, Leu2-Gly77
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS with 5% trehalose
          Contains
          no preservative
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3702112
          Product Specific Information
          Reconstitute at 0.5 mg/mL in sterile PBS.
          Target Information
          NEDD8 is a ubiquitin-like protein which plays an important role in cell cycle control and embryogenesis. Covalent attachment to its substrates requires prior activation by the E1 complex UBE1C-APPBP1 and linkage to the E2 enzyme UBE2M. Attachment of NEDD8 to cullins activates their associated E3 ubiquitin ligase activity, and thus promotes polyubiquitination and proteasomal degradation of cyclins and other regulatory proteins.
          Bioinformatics
          Protein Aliases: NEDD-8; NEDD8; Neddylin; Neural precursor cell expressed developmentally down-regulated protein 8; neural precursor cell expressed, developmentally down-regulated 8; neural precursor cell expressed, developmentally down-regulated gene 8; Ubiquitin-like protein NEDD8
          View more View less
          Gene Aliases: NEDD-8; NEDD8; Rub1
          View more View less
          UniProt ID: (Human) Q15843, (Rat) Q71UE8, (Mouse) P29595
          View more View less
          Entrez Gene ID: (Human) 4738, (Rat) 25490, (Mouse) 18002
          View more View less
          Function(s)
          protein binding protein tag ubiquitin protein ligase binding
          Process(es)
          regulation of transcription from RNA polymerase II promoter protein localization response to organic cyclic compound protein neddylation proteolysis ubiquitin-dependent protein catabolic process anatomical structure morphogenesis protein ubiquitination modification-dependent protein catabolic process regulation of proteolysis protein modification process cellular response to camptothecin regulation of postsynapse assembly
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline