    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Caspase 10 Antibodies
          Invitrogen
          Caspase 10 Monoclonal Antibody (63131)
          View all (14) Caspase 10 antibodies
          Questions & Answers
          Cite Caspase 10 Monoclonal Antibody (63131)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Caspase 10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Caspase 10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Caspase 10 Antibody (MA5-61938) in WB
          Western Blot analysis of Caspase 10 in Jurkat using Caspase 10 monoclonal antibody (Product # MA5-61938). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Caspase 10 Antibody in Western Blot (WB)
          Caspase 10 Monoclonal Antibody (63131)
          Product Details
          MA5-61938
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          63131
          Immunogen
          E. coli-derived recombinant human Caspase-10/a/B (Mch 4), Val220-Ile479
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS with 5% trehalose
          Contains
          no preservative
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3702782
          Product Specific Information
          Reconstitute at 0.5 mg/mL in sterile PBS.
          Target Information
          This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein cleaves and activates caspases 3 and 7, and the protein itself is processed by caspase 8. Mutations in this gene are associated with apoptosis defects seen in type II autoimmune lymphoproliferative syndrome. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
          Bioinformatics
          Protein Aliases: Apoptotic protease Mch-4; CASP-10; caspase 10 apoptosis-related cysteine peptidase; caspase 10, apoptosis-related cysteine protease; Caspase-10; FADD-like ICE2; FAS-associated death domain protein interleukin-1B-converting enzyme 2; FLICE2; FLICE2; ICE like protease; caspase-10/b; ICE-like apoptotic protease 4; ICE-like protease; interleukin-1B-converting enzyme 2; unnamed protein product
          View more View less
          Gene Aliases: ALPS2; CASP10; FLICE-2; FLICE2; MCH4
          View more View less
          UniProt ID: (Human) Q92851
          View more View less
          Entrez Gene ID: (Human) 843
          View more View less
          Function(s)
          cysteine-type endopeptidase activity protein binding peptidase activity cysteine-type peptidase activity hydrolase activity ubiquitin protein ligase binding death effector domain binding
          Process(es)
          proteolysis apoptotic process regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of neuron apoptotic process protein maturation execution phase of apoptosis positive regulation of execution phase of apoptosis
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline