    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • VSIG8 Antibodies
          Invitrogen
          VSIG8 Monoclonal Antibody (961823)
          View all (7) VSIG8 antibodies
          Questions & Answers
          Cite VSIG8 Monoclonal Antibody (961823)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          VSIG8 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          VSIG8 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          VSIG8 Antibody (MA5-62025) in Flow
          Flow Cytometry analysis of VSIG8 in HEK293 Human Cell Line using VSIG8 monoclonal antibody (Product # MA5-62025). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          VSIG8 Antibody in Flow Cytometry (Flow)
          VSIG8 Monoclonal Antibody (961823)
          Product Details
          MA5-62025
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          961823
          Immunogen
          Human embryonic kidney cell line HEK293-derived transfected with human VSIG8, Met1-Gly263
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 5% trehalose
          Contains
          no preservative
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3702957
          Product Specific Information
          Reconstitute at 0.5 mg/mL in sterile PBS.
          Target Information
          VSIG8 contains 2 Ig-like V-type (immunoglobulin-like) domains. VSIG8 is single-pass type I membrane protein. The function of the VSIG8 protein remains unknown.
          Bioinformatics
          Protein Aliases: V-set and immunoglobulin domain-containing protein 8
          View more View less
          Gene Aliases: C1orf204; VSIG8
          View more View less
          UniProt ID: (Human) P0DPA2
          View more View less
          Entrez Gene ID: (Human) 391123
          View more View less
          Function(s)
          RNA binding protein binding
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline