    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CDK1 Antibodies
          Invitrogen
          CDK1 Monoclonal Antibody (POH-1), Alexa Fluor™ 488
          Greener Choice
          View all (91) CDK1 antibodies
          Questions & Answers
          Cite CDK1 Monoclonal Antibody (POH-1), Alexa Fluor™ 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CDK1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CDK1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CDK1 Antibody (MA562364) in ICC/IF
          Immunocytochemistry analysis of CDK1 in HeLa cells. Samples were incubated with CDK1 monoclonal antibody (Product # MA562364) using a dilution of 2 µg/mL (green). Cell nuclei stained with DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CDK1 Antibody in Immunocytochemistry (ICC/IF)
          CDK1 Monoclonal Antibody (POH-1), Alexa Fluor™ 488
          Product Details
          MA562364
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          2 µg/mL
          -
          Product Specifications
          Species Reactivity
          Bovine, Human, Non-human primate
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          POH-1
          Immunogen
          Bacterially expressed full-length human Cdk1 protein.
          3D Epitope / Immunogen
          Conjugate
          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          499/520 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3703550
          Target Information
          CDK1 or Cell Division Control protein 1 is essential for the completion of START, the controlling event in the cell cycle that is required to initiate mitosis. CDK1 is a catalytic subunit of a protein kinase complex, called the M-Phase Promoting Factor that induces entry into mitosis and is universal among eukaryotes. Phosphorylation of Bcl-2 in G2/M phase-arrested cells following photodynamic therapy with hypericin involves a CDK1-mediated signal and delays the onset of apoptosis. Therapeutic potential of CDK inhibitor NU2058 in androgen-independent prostate cancer has also been demonstrated.
          Bioinformatics
          Protein Aliases: ABL1; CDK1; cell cycle controller CDC2; Cell division control protein 2 homolog; cell division cycle 2, G1 to S and G2 to M; Cell division protein kinase 1; Cyclin-dependent kinase 1; DKFZp686L20222; KCIP-1; MGC111195; MGC53829; OTTHUMP00000019660; P34 CDK1; P34 MGC111195; p34 protein kinase; Protein 1054; unnamed protein product
          View more View less
          Gene Aliases: CDC2; CDC28A; CDK1; CDKN1; P34CDC2
          View more View less
          UniProt ID: (Bovine) P48734, (Human) P06493
          View more View less
          Entrez Gene ID: (Bovine) 281061, (Human) 983
          View more View less
          Function(s)
          chromatin binding cyclin-dependent protein serine/threonine kinase activity ATP binding cyclin binding Hsp70 protein binding histone kinase activity nucleotide binding virus receptor activity protein kinase activity protein serine/threonine kinase activity protein binding RNA polymerase II carboxy-terminal domain kinase activity kinase activity transferase activity cyclin-dependent protein kinase activity protein serine kinase activity non-receptor serine/threonine protein kinase
          Process(es)
          apoptotic process cell proliferation positive regulation of G2/M transition of mitotic cell cycle histone phosphorylation peptidyl-serine phosphorylation peptidyl-threonine phosphorylation epithelial cell differentiation protein localization to kinetochore regulation of circadian rhythm negative regulation of apoptotic process rhythmic process cell division Golgi disassembly positive regulation of protein localization to nucleus positive regulation of mitochondrial ATP synthesis coupled electron transport G1/S transition of mitotic cell cycle G2/M transition of mitotic cell cycle microtubule cytoskeleton organization DNA replication DNA repair chromatin remodeling regulation of transcription from RNA polymerase II promoter cellular response to DNA damage stimulus nuclear envelope organization mitotic nuclear envelope disassembly mitotic G2 DNA damage checkpoint centrosome cycle pronuclear fusion response to toxic substance positive regulation of gene expression negative regulation of gene expression regulation of Schwann cell differentiation response to amine response to activity cell migration protein deubiquitination chromosome condensation positive regulation of protein import into nucleus response to hydrogen peroxide mitotic cell cycle phase transition response to ethanol positive regulation of DNA replication regulation of embryonic development thymidine biosynthetic process response to cadmium ion response to copper ion viral entry into host cell microtubule polymerization fibroblast proliferation response to axon injury ventricular cardiac muscle cell development positive regulation of cardiac muscle cell proliferation cilium disassembly positive regulation of mitotic sister chromatid segregation protein-containing complex assembly cellular response to hydrogen peroxide ERK1 and ERK2 cascade DNA strand resection involved in replication fork processing regulation of mitotic attachment of spindle microtubules to kinetochore microtubule cytoskeleton organization involved in mitosis DNA synthesis involved in mitotic DNA replication protein localization to site of double-strand break mitotic DNA-dependent DNA replication
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline