    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GYS1 Antibodies
          Invitrogen
          GYS1 Recombinant Rabbit Monoclonal Antibody (K01_1R80)
          View all (32) GYS1 antibodies
          Questions & Answers
          Cite GYS1 Recombinant Rabbit Monoclonal Antibody (K01_1R80)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          GYS1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GYS1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GYS1 Antibody (MA563572) in ICC/IF
          Immunocytochemistry analysis of GYS1 (green) in HeLa cells. Samples were incubated with GYS1 recombinant monoclonal (Product # MA563572). DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GYS1 Antibody in Immunocytochemistry (ICC/IF)
          GYS1 Antibody in Western Blot (WB)
          GYS1 Antibody in Western Blot (WB)
          Product Details
          MA563572
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_1R80
          Immunogen
          A synthetic peptide of human Glycogen synthase 1.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          50mM tris glycine, pH 7.4, with 40% glycerol, 0.05% BSA, 150mM NaCl
          Contains
          0.01% sodium azide
          Storage conditions
          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3704620
          Target Information
          Glycogen Synthase 1 (GYS1) catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: EC 2.4.1.11; Glycogen [starch] synthase, muscle; glycogen synthase; Glycogen synthase 1; glycogen synthase 1 (muscle); unnamed protein product
          View more View less
          Gene Aliases: GSY; GYS; GYS1
          View more View less
          UniProt ID: (Human) P13807, (Rat) A2RRU1
          View more View less
          Entrez Gene ID: (Human) 2997, (Rat) 690987
          View more View less
          Function(s)
          catalytic activity glycogen (starch) synthase activity protein binding glucose binding transferase activity transferase activity, transferring glycosyl groups glycogen synthase activity, transferring glucose-1-phosphate protein kinase binding
          Process(es)
          glycogen metabolic process glycogen biosynthetic process heart development
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline