    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SNX4 Antibodies
          Invitrogen
          SNX4 Recombinant Rabbit Monoclonal Antibody (K01_1Y62)
          View all (16) SNX4 antibodies
          Questions & Answers
          Cite SNX4 Recombinant Rabbit Monoclonal Antibody (K01_1Y62)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          SNX4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SNX4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SNX4 Antibody (MA563619) in WB
          Western blot analysis of SNX4 in various lysates. Samples were incubated with SNX4 recombinant monoclonal (Product # MA563619). Samples: C6, 3T3, Hela lysates. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SNX4 Antibody in Western Blot (WB)
          Product Details
          MA563619
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_1Y62
          Immunogen
          A synthetic peptide of human SNX4.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          50mM tris glycine, pH 7.4, with 40% glycerol, 150mM NaCl, 0.05% BSA
          Contains
          0.01% sodium azide
          Storage conditions
          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3704667
          Target Information
          SNX2 and SNX4 share a high degree of amino acid similarity with SNX1, as they all contain a characteristic phox homology (PX) domain. These proteins are all partially associated with cellular membranes, and they, likewise, associate with EGF, PDGF and insulin receptor tyrosine kinases. These nexins are widely expressed and yet have various tissue distribution patterns. Additionally, the sorting nexins can associate with each other and with a variety of other cellular proteins, suggesting that they exist as part of multisubunit complexes. The related protein, SNX3, comprises a distinct subgroup of nexins that share less sequence similarity outside of the PX domain and have dramatically different binding affinities for the tyrosine kinase receptors.
          Bioinformatics
          Protein Aliases: SNX4; Sorting nexin-4; unnamed protein product
          View more View less
          Gene Aliases: 1810036H14Rik; AI037066; ATG24B; SNX4
          View more View less
          UniProt ID: (Human) O95219, (Mouse) Q91YJ2
          View more View less
          Entrez Gene ID: (Human) 8723, (Mouse) 69150, (Rat) 360725
          View more View less
          Function(s)
          epidermal growth factor receptor binding insulin receptor binding protein binding lipid binding phosphatidylinositol-3-phosphate binding phosphatidylinositol binding transferrin receptor binding leptin receptor binding scaffold/adaptor protein
          Process(es)
          protein transport endocytic recycling positive regulation of histamine secretion by mast cell positive regulation of autophagosome assembly transport endocytosis vesicle organization
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline