    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MLK3 Antibodies
          Invitrogen
          MLK3 Recombinant Rabbit Monoclonal Antibody (K01_1T96)
          View all (28) MLK3 antibodies
          Questions & Answers
          Cite MLK3 Recombinant Rabbit Monoclonal Antibody (K01_1T96)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          MLK3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MLK3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          MLK3 Antibody (MA563823) in WB
          Western blot analysis of MLK3 in various lysates. Samples were incubated with MLK3 recombinant monoclonal (Product # MA563823). Samples: HepG2, A431 lysates. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MLK3 Antibody in Western Blot (WB)
          Product Details
          MA563823
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_1T96
          Immunogen
          A synthetic peptide of human MLK3.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          50mM tris glycine, pH 7.4, with 40% glycerol, 0.05% BSA, 150mM NaCl
          Contains
          0.01% sodium azide
          Storage conditions
          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3704871
          Target Information
          MAP3K11 (MLK3) is a serine/threonine kinase that activates both the p38 MAP kinase and c-Jun kinase pathways. This kinase contains a SH3 domain and a leucine zipper-basic motif. This kinase preferentially activates MAPK8/JNK kinase, and functions as a positive regulator of JNK signaling pathway. This kinase can directly phosphorylate, and activates IkappaB kinase alpha and beta, and is found to be involved in the transcription activity of NF-kappaB mediated by Rho family GTPases and CDC42.
          Bioinformatics
          Protein Aliases: MGC17114; Mitogen-activated protein kinase kinase kinase 11; Mixed lineage kinase 3; protein-tyrosine kinase PTK1; SH3 domain-containing proline-rich kinase; Src-homology 3 domain-containing proline-rich kinase; unnamed protein product
          View more View less
          Gene Aliases: MAP3K11; MEKK11; MLK-3; MLK3; PTK1; SPRK
          View more View less
          UniProt ID: (Human) Q16584
          View more View less
          Entrez Gene ID: (Human) 4296
          View more View less
          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity JUN kinase kinase kinase activity MAP kinase kinase kinase activity protein binding ATP binding kinase activity transferase activity small GTPase binding mitogen-activated protein kinase kinase binding mitogen-activated protein kinase kinase kinase binding identical protein binding protein homodimerization activity protein serine kinase activity
          Process(es)
          MAPK cascade protein phosphorylation microtubule-based process JNK cascade positive regulation of apoptotic process positive regulation of JUN kinase activity positive regulation of neuron apoptotic process cell cycle G1/S phase transition positive regulation of JNK cascade protein autophosphorylation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline